Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00813333_m1
          See other TRMT1 GE Assays ›
          Gene Symbol:
          Trmt1
          Gene Name:
          tRNA methyltransferase 1
          Gene Aliases:
          6720406L13Rik, 6720477L24, D8Ertd812e
          Chromosome Location:
          Chr.8: 84688725 - 84699808 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00813333_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trmt1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164559.1 NP_001158031.1 5 - 6 761 62
          NM_001164560.1 NP_001158032.1 5 - 6 685 62
          NM_198020.2 NP_932137.2 5 - 6 788 62
          GenBank mRNA AK032945.1 - 4 - 5 934 62
          AK078395.1 - 4 - 5 964 62
          AK084775.1 - 5 - 6 726 62
          AK155251.1 - 5 - 6 753 62
          AK159467.1 - 5 - 6 726 62
          AK170412.1 - 5 - 6 650 62
          AK171002.1 - 5 - 6 753 62
          BC027404.1 - 5 - 6 757 4950980 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800003233.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Trmt1

          Panther Classification:

          Molecular Function -

          RNA methyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA N2-guanine methylation
          tRNA processing
          biological_process
          methylation
          tRNA binding
          RNA binding
          tRNA (guanine-N2-)-methyltransferase activity
          methyltransferase activity
          transferase activity
          poly(A) RNA binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0800007212.mm.1 NM_001164559.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007212.mm.1 NM_001164559.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007212.mm.1 NM_001164560.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007212.mm.1 NM_001164560.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007212.mm.1 NM_198020.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007212.mm.1 NM_198020.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007212.mm.1 TC0800003233.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800007216.mm.1 NM_001164559.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007216.mm.1 NM_001164559.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007216.mm.1 NM_001164560.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007216.mm.1 NM_001164560.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007216.mm.1 NM_198020.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007216.mm.1 NM_198020.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007216.mm.1 TC0800003233.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-d59m7:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0