Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00824350_m1
          See other VMN2R122 GE Assays ›
          Gene Symbol:
          Vmn2r122
          Gene Name:
          vomeronasal 2, receptor, 122
          Gene Aliases:
          V2r12, V2r2, Vmn2r-ps158
          Chromosome Location:
          Chr.4|NT_187052: 3536 - 11935 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00824350_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn2r122
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009488.2 NP_033514.2 4 - 5 1503 98
          GenBank mRNA AF011422.1 - 4 - 5 1478 98

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn2r122

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09579794 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09579783 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09579773 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09579794 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10405871 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410206 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373688 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410205 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373685 - -
          Affymetrix Mouse Gene 1.1 ST Array 10373684 - -
          Affymetrix Mouse Gene 1.1 ST Array 10598118 - -
          Affymetrix Mouse Gene 1.1 ST Array 10405870 - -
          Affymetrix Mouse Gene 1.1 ST Array 10410204 - -
          Affymetrix Mouse Clariom S Array PSR4_JH584292_random00000010.mm.1 NM_009488.2 Refseq
          Affymetrix Mouse Clariom S Array PSR4_JH584292_random00000010.mm.1 NM_009492.2 Refseq
          Affymetrix Mouse Clariom S Array PSR4_JH584292_random00000010.mm.1 TC4_JH584292_random00000001.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR4_JH584292_random00000004.mm.1 NM_009488.2 Refseq
          Affymetrix Mouse Clariom S Array PSR4_JH584292_random00000004.mm.1 NM_009492.2 Refseq
          Affymetrix Mouse Clariom S Array PSR4_JH584292_random00000004.mm.1 TC4_JH584292_random00000001.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR4_JH584292_random00000011.mm.1 NM_009488.2 Refseq
          Affymetrix Mouse Clariom S Array PSR4_JH584292_random00000011.mm.1 NM_009492.2 Refseq
          Affymetrix Mouse Clariom S Array PSR4_JH584292_random00000011.mm.1 TC4_JH584292_random00000001.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR4_JH584292_random00000003.mm.1 NM_009488.2 Refseq
          Affymetrix Mouse Clariom S Array PSR4_JH584292_random00000003.mm.1 NM_009492.2 Refseq
          Affymetrix Mouse Clariom S Array PSR4_JH584292_random00000003.mm.1 TC4_JH584292_random00000001.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR4_JH584292_random00000005.mm.1 NM_009488.2 Refseq
          Affymetrix Mouse Clariom S Array PSR4_JH584292_random00000005.mm.1 NM_009492.2 Refseq
          Affymetrix Mouse Clariom S Array PSR4_JH584292_random00000005.mm.1 TC4_JH584292_random00000001.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR4_JH584292_random00000002.mm.1 NM_009488.2 Refseq
          Affymetrix Mouse Clariom S Array PSR4_JH584292_random00000002.mm.1 NM_009492.2 Refseq
          Affymetrix Mouse Clariom S Array PSR4_JH584292_random00000002.mm.1 TC4_JH584292_random00000001.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR4_JH584292_random00000006.mm.1 NM_009488.2 Refseq
          Affymetrix Mouse Clariom S Array PSR4_JH584292_random00000006.mm.1 NM_009492.2 Refseq
          Affymetrix Mouse Clariom S Array PSR4_JH584292_random00000006.mm.1 TC4_JH584292_random00000001.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR4_JH584292_random00000008.mm.1 NM_009488.2 Refseq
          Affymetrix Mouse Clariom S Array PSR4_JH584292_random00000008.mm.1 TC4_JH584292_random00000001.mm.1 MTA TC
          Murine Genome U74v2 Set 95798_PM_f_at - -
          Murine Genome U74v2 Set 101644_PM_f_at - -
          Murine Genome U74v2 Set 100677_f_at - -
          Murine Genome U74v2 Set 101630_PM_f_at - -
          Murine Genome U74v2 Set 101630_f_at - -
          Murine Genome U74v2 Set 101644_f_at - -
          Murine Genome U74v2 Set 95798_f_at - -
          Murine Genome U74v2 Set 100677_PM_f_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1240064 - -
          MouseWG-6 v2 Expression BeadChip 2260672 - -
          Illumina mouse-6 v1.0 expression beadchip 1240064 - -
          Illumina mouse-6 v1.0 expression beadchip 430487 - -
          Illumina mouse-6 v1.0 expression beadchip 2260672 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001240064 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002260672 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000430487 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2478483 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2455694 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2455694 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2478483 - -
          mouse-6 v1.1 (Illumina) scl022303.3_56-S - -
          mouse-6 v1.1 (Illumina) scl0022303.1_104-S - -
          mouse-6 v1.1 (Illumina) scl0022303.1_124-S - -
          Illumina Sentrix mouse-6.v1 bead array 2260672 - -
          Illumina Sentrix mouse-6.v1 bead array 1240064 - -
          Illumina Sentrix mouse-6.v1 bead array 430487 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0022303.1_104-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl022303.3_56-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0022303.1_124-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2260672 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 430487 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1240064 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2478483 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2500450 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2478483 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2500445 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0022303.1_104-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline