Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00833443_s1
          See other IFNAB GE Assays ›
          Gene Symbol:
          Ifnab
          Gene Name:
          interferon alpha B
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 88690599 - 88691268 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00833443_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ifnab
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008336.2 NP_032362.2 1 384 230
          GenBank mRNA BC116862.1 - 1 384 40126181 230
          BC116864.1 - 1 384 40126183 230

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ifnab

          Panther Classification:

          Molecular Function -

          interferon superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          adaptive immune response
          T cell activation involved in immune response
          natural killer cell activation involved in immune response
          humoral immune response
          cytokine-mediated signaling pathway
          B cell differentiation
          positive regulation of peptidyl-serine phosphorylation of STAT protein
          B cell proliferation
          response to exogenous dsRNA
          cytokine activity
          type I interferon receptor binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030077 NM_008336.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030077 BC116862.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030077 BC116864.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030076 NM_008336.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030076 BC116862.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030076 BC116864.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030076 NM_008336.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030076 BC116862.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030076 BC116864.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030077 NM_008336.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030077 BC116862.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030077 BC116864.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P387681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091722 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091720 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091705 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091711 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091713 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091708 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091703 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091699 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091714 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091717 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091709 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091712 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091694 - -
          Whole Mouse Genome G4122A A_51_P387681 - -
          Whole Mouse Genome G4122A A_51_P465436 BC116862.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P465436 BC116864.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03716407 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03716392 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03716398 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03716397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091738 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03729041 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091723 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091711 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03933136 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091708 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091683 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091705 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091698 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03933129 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03933137 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091707 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091721 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091726 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03762198 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03716400 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03762203 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091740 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091729 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10514291 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514290 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514318 - -
          Affymetrix HT MG-430 PM Array Plate 1450613_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10514317 - -
          Mouse Genome 430 2.0 Array 1450613_x_at - -
          Mouse Genome 430 2.0 Array 1450613_PM_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15974_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15974_at - -
          Mouse Expression Set 430 1450593_at - -
          Mouse Expression Set 430 1422404_PM_x_at - -
          Mouse Expression Set 430 1422403_PM_at - -
          Mouse Expression Set 430 1450593_PM_at - -
          Mouse Expression Set 430 1422403_at - -
          Mouse Expression Set 430 1450613_PM_x_at - -
          Mouse Expression Set 430 1422404_x_at - -
          Mouse Expression Set 430 1450613_x_at - -
          Murine Genome U74v2 Set 101317_PM_f_at - -
          Murine Genome U74v2 Set 96419_PM_f_at - -
          Murine Genome U74v2 Set 101619_at - -
          Murine Genome U74v2 Set 102149_f_at - -
          Murine Genome U74v2 Set 101619_PM_at - -
          Murine Genome U74v2 Set 96419_f_at - -
          Murine Genome U74v2 Set 102149_PM_f_at - -
          Murine Genome U74v2 Set 101317_f_at - -
          Mouse Genome 430A 2.0 Array 1450593_PM_at - -
          Mouse Genome 430A 2.0 Array 1422403_PM_at - -
          Mouse Genome 430A 2.0 Array 1450613_x_at - -
          Mouse Genome 430A 2.0 Array 1422404_x_at - -
          Mouse Genome 430A 2.0 Array 1450613_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1422404_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1422403_at - -
          Mouse Genome 430A 2.0 Array 1450593_at - -
          Affymetrix HT Mouse Genome 430A Array 1450613_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1450613_x_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103520288 - -
          Illumina mouse-6 v1.0 expression beadchip 103520288 - -
          Illumina mouse-6 v1.0 expression beadchip 2030687 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002030687 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2770778 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2770778 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1223579 - -
          mouse-6 v1.1 (Illumina) scl015974.1_123-S - -
          mouse-6 v1.1 (Illumina) GI_6680370-S - -
          Illumina Sentrix mouse-6.v1 bead array 2030687 - -
          Illumina Sentrix mouse-6.v1 bead array 103520288 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl015974.1_123-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103520288 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2030687 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2770778 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1223579 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2770778 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline