Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00833737_mH
          See other VMN2R-PS159 GE Assays ›
          Gene Symbol:
          Vmn2r-ps159
          Gene Name:
          vomeronasal 2, receptor, pseudogene 159
          Gene Aliases:
          V2r8, V2r9, Vmn2r-ps160
          Chromosome Location:
          Chr.4: 156331103 - 156339499 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00833737_mH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn2r-ps159
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038052.1 2 - 3 1092 96
          GenBank mRNA AF011418.1 - 2 - 3 1092 96

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          MicroArray Probes:

          Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10608088 - -
          Affymetrix Mouse Gene 1.0 ST Array 10405871 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606560 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373688 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414574 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373685 - -
          Affymetrix Mouse Gene 1.1 ST Array 10414561 - -
          Affymetrix Mouse Gene 1.1 ST Array 10373684 - -
          Affymetrix Mouse Gene 1.1 ST Array 10606559 - -
          Affymetrix Mouse Gene 1.1 ST Array 10405870 - -
          Affymetrix Mouse Gene 1.1 ST Array 10608087 - -
          Affymetrix Mouse Gene 1.1 ST Array 10598118 - -
          Mouse Genome 430 2.0 Array 1450338_x_at - -
          Mouse Genome 430 2.0 Array 1450338_PM_x_at - -
          Affymetrix Mouse Clariom S Array PSR0400017855.mm.1 NR_038052.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400017855.mm.1 TC0400002112.mm.1 MTA TC
          Mouse Expression Set 430 1450338_x_at - -
          Mouse Expression Set 430 1450338_PM_x_at - -
          Murine Genome U74v2 Set 95798_f_at - -
          Murine Genome U74v2 Set 100677_PM_f_at - -
          Murine Genome U74v2 Set 100677_f_at - -
          Murine Genome U74v2 Set 101630_f_at - -
          Murine Genome U74v2 Set 101630_PM_f_at - -
          Murine Genome U74v2 Set 101644_f_at - -
          Murine Genome U74v2 Set 101644_PM_f_at - -
          Murine Genome U74v2 Set 95798_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1450338_x_at - -
          Mouse Genome 430A 2.0 Array 1450338_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 352192 - -
          ABI Mouse Genome Survey Microarray 716624 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2690100 - -
          Illumina mouse-6 v1.0 expression beadchip 2690100 - -
          Illumina mouse-6 v1.0 expression beadchip 3710433 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003710433 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002690100 - -
          mouse-6 v1.1 (Illumina) scl0022314.1_235-S - -
          mouse-6 v1.1 (Illumina) scl022314.3_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 2690100 - -
          Illumina Sentrix mouse-6.v1 bead array 3710433 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl022314.3_0-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0022314.1_235-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3710433 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2690100 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2992339 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2455694 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2992339 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2455694 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline