Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00833923_m1
          See other GSTM3 GE Assays ›
          Gene Symbol:
          Gstm3
          Gene Name:
          glutathione S-transferase, mu 3
          Gene Aliases:
          Fsc2, mGSTM5
          Chromosome Location:
          Chr.3: 107963696 - 107969174 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00833923_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gstm3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010359.2 NP_034489.1 7 - 8 602 155
          GenBank mRNA BC044927.1 - 7 - 8 586 5051815 155
          J03953.1 - 6 - 7 542 155

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300003246.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gstm3

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          cellular response to drug
          cellular response to organic cyclic compound
          glutathione transferase activity
          transferase activity
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2102060 NM_010359.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2102060 BC044927.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2065231 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 212569 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 738608 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 48036 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 126040 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101287 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174078 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173576 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2065231 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2102060 NM_010359.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2102060 BC044927.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P454949 J03953.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22409828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22409832 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22409853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22409831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22409840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22409844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22409803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22409805 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22409813 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22409816 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22409845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22409848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22409815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22409833 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22409855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22409850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22409821 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22409858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22409857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22409806 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22409808 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22409854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22409817 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22409846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22409823 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22409842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22409837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22409827 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22409826 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P173576 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101287 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174078 - -
          Whole Mouse Genome G4122A A_51_P454949 J03953.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174078 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101287 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P173576 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03166337 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03166339 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03166336 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03166352 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03166334 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03166353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22409853 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409808 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409864 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409778 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03166337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02787426 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409787 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409858 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02771760 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409777 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02746842 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409833 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409772 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02682685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409802 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409863 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409821 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409789 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409848 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409819 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03143730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409794 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409779 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409840 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02837431 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03049877 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409814 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409827 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409861 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409806 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409832 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409722 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409831 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02356090 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409791 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409770 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409826 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409782 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409786 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409846 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03088443 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02746845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02319026 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409769 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02553874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409724 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02356094 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409823 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409842 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409853 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22409860 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10362512 - -
          Affymetrix Mouse Gene 1.0 ST Array 10501221 - -
          Affymetrix Mouse Gene 1.0 ST Array 10501219 - -
          Affymetrix Mouse Gene 1.0 ST Array 10501220 - -
          Affymetrix HT MG-430 PM Array Plate 1427474_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1427473_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10501218 - -
          Affymetrix Mouse Gene 1.1 ST Array 10362511 - -
          Mouse Genome 430 2.0 Array 1427473_PM_at - -
          Mouse Genome 430 2.0 Array 1427473_at - -
          Mouse Genome 430 2.0 Array 1427474_PM_s_at - -
          Mouse Genome 430 2.0 Array 1427474_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14864_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14864_at - -
          Affymetrix Mouse Clariom S Array PSR0300021293.mm.1 NM_010359.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300021293.mm.1 NM_010359.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300021293.mm.1 TC0300003246.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300021294.mm.1 NM_010359.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300021294.mm.1 NM_010359.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300021294.mm.1 TC0300003246.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300021295.mm.1 NM_010359.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300021295.mm.1 NM_010359.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300021295.mm.1 TC0300003246.mm.1 MTA TC
          Mouse Expression Set 430 1427474_PM_s_at - -
          Mouse Expression Set 430 1427473_at - -
          Mouse Expression Set 430 1427473_PM_at - -
          Mouse Expression Set 430 1427474_s_at - -
          Murine Genome U74v2 Set 97682_PM_r_at - -
          Murine Genome U74v2 Set 97681_PM_f_at - -
          Murine Genome U74v2 Set 97681_f_at - -
          Murine Genome U74v2 Set 97682_r_at - -
          Mouse Genome 430A 2.0 Array 1427473_at - -
          Mouse Genome 430A 2.0 Array 1427474_s_at - -
          Mouse Genome 430A 2.0 Array 1427473_PM_at - -
          Mouse Genome 430A 2.0 Array 1427474_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1427473_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427473_at - -
          Affymetrix HT Mouse Genome 430A Array 1427474_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1427474_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 665168 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2570451 - -
          Illumina mouse-6 v1.0 expression beadchip 101170047 - -
          Illumina mouse-6 v1.0 expression beadchip 2570451 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002570451 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1216149 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1219393 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1216149 - -
          mouse-6 v1.1 (Illumina) scl014864.1_330-S - -
          mouse-6 v1.1 (Illumina) GI_33468898-S - -
          Illumina Sentrix mouse-6.v1 bead array 101170047 - -
          Illumina Sentrix mouse-6.v1 bead array 2570451 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl014864.1_330-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101170047 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2570451 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1216149 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1216149 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline