Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00833983_s1
          See other IFNA9 GE Assays ›
          Gene Symbol:
          Ifna9
          Gene Name:
          interferon alpha 9
          Gene Aliases:
          Ifa9
          Chromosome Location:
          Chr.4: 88591813 - 88592385 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00833983_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ifna9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010507.1 NP_034637.1 1 438 193
          GenBank mRNA BC117775.1 - 1 433 193

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003002.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ifna9

          Panther Classification:

          Molecular Function -

          interferon superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          adaptive immune response
          T cell activation involved in immune response
          natural killer cell activation involved in immune response
          defense response
          humoral immune response
          cytokine-mediated signaling pathway
          B cell differentiation
          positive regulation of peptidyl-serine phosphorylation of STAT protein
          B cell proliferation
          response to exogenous dsRNA
          defense response to virus
          cytokine activity
          cytokine receptor binding
          type I interferon receptor binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P363308 NM_010507.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P363308 BC117775.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091307 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091315 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091299 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091288 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091302 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091310 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091295 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091287 - -
          Whole Mouse Genome G4122A A_51_P363308 NM_010507.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P363308 BC117775.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114071 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091315 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03548890 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03659836 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091288 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091287 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03548891 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03309758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091339 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091275 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03334001 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091278 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03535023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03846618 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091306 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091285 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091296 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03548889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091303 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091299 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091316 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091319 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091297 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03529273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091315 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091314 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03668268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03309757 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091312 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091332 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10505880 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514291 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514297 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514290 - -
          Affymetrix HT MG-430 PM Array Plate 1422406_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10514296 - -
          Mouse Genome 430 2.0 Array 1422406_PM_at - -
          Mouse Genome 430 2.0 Array 1422406_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15972_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15972_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400024296.mm.1 TC0400003002.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400024296.mm.1 NM_010507.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400024296.mm.1 NM_010507.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400024297.mm.1 TC0400003002.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400024297.mm.1 NM_010507.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400024297.mm.1 NM_010507.1 Refseq
          Mouse Expression Set 430 1422403_PM_at - -
          Mouse Expression Set 430 1422404_PM_x_at - -
          Mouse Expression Set 430 1450593_at - -
          Mouse Expression Set 430 1422406_PM_at - -
          Mouse Expression Set 430 1450593_PM_at - -
          Mouse Expression Set 430 1422403_at - -
          Mouse Expression Set 430 1422404_x_at - -
          Mouse Expression Set 430 1422406_at - -
          Mouse Expression Set 430 1460611_PM_at - -
          Mouse Expression Set 430 1460611_at - -
          Murine Genome U74v2 Set 101619_at - -
          Murine Genome U74v2 Set 101788_f_at - -
          Murine Genome U74v2 Set 102149_PM_f_at - -
          Murine Genome U74v2 Set 96419_PM_f_at - -
          Murine Genome U74v2 Set 96419_f_at - -
          Murine Genome U74v2 Set 101788_PM_f_at - -
          Murine Genome U74v2 Set 102148_f_at - -
          Murine Genome U74v2 Set 102148_PM_f_at - -
          Murine Genome U74v2 Set 101619_PM_at - -
          Murine Genome U74v2 Set 102149_f_at - -
          Mouse Genome 430A 2.0 Array 1422403_PM_at - -
          Mouse Genome 430A 2.0 Array 1450593_at - -
          Mouse Genome 430A 2.0 Array 1422404_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1422404_x_at - -
          Mouse Genome 430A 2.0 Array 1450593_PM_at - -
          Mouse Genome 430A 2.0 Array 1422406_at - -
          Mouse Genome 430A 2.0 Array 1422403_at - -
          Mouse Genome 430A 2.0 Array 1422406_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4010372 - -
          Illumina mouse-6 v1.0 expression beadchip 105720138 - -
          Illumina mouse-6 v1.0 expression beadchip 4010372 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004010372 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2648399 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2750971 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1237080 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2750971 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2648399 - -
          mouse-6 v1.1 (Illumina) GI_6754297-S - -
          mouse-6 v1.1 (Illumina) scl015972.1_202-S - -
          Illumina Sentrix mouse-6.v1 bead array 4010372 - -
          Illumina Sentrix mouse-6.v1 bead array 105720138 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl015972.1_202-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105720138 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4010372 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2750971 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2750971 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1237080 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0