Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00834384_g1
          See other HSPB1 GE Assays ›
          Gene Symbol:
          Hspb1
          Gene Name:
          heat shock protein 1
          Gene Aliases:
          27kDa, Hsp25
          Chromosome Location:
          Chr.5: 135887919 - 135889563 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00834384_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hspb1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013560.2 NP_038588.2 2 - 3 578 171
          GenBank mRNA AK003119.1 - 2 - 3 499 171
          AK010663.1 - 2 - 3 496 171
          AK168927.1 - 2 - 3 496 171
          BC018257.1 - 2 - 3 518 3498180 171
          BC099463.1 - 2 - 3 473 30848584 171
          U03560.1 - 2 - 3 435 171
          U03561.1 - 3 - 4 402 171
          U03562.1 - 3 - 4 336 171
          X14686.1 - 2 - 3 480 171
          X14687.1 - 2 - 3 525 171

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hspb1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of protein kinase activity
          response to unfolded protein
          positive regulation of interleukin-1 beta production
          intracellular signal transduction
          cellular response to vascular endothelial growth factor stimulus
          positive regulation of endothelial cell chemotaxis by VEGF-activated vascular endothelial growth factor receptor signaling pathway
          positive regulation of tumor necrosis factor biosynthetic process
          regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of blood vessel endothelial cell migration
          positive regulation of angiogenesis
          platelet aggregation
          negative regulation of protein serine/threonine kinase activity
          negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
          positive regulation of endothelial cell chemotaxis
          negative regulation of apoptotic signaling pathway
          protein kinase C binding
          protein kinase C inhibitor activity
          protein kinase binding
          identical protein binding
          ubiquitin binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24118365 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039284 X14686.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039284 AK010663.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039284 AK168927.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039284 AV458710.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039284 BC018257.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 825891 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039284 X14686.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039284 AK010663.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039284 AK168927.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039284 AV458710.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039284 BC018257.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24118349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24118354 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24118376 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24118347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24118361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24118359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24118345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24118351 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24118367 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24118371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24118355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24118377 - -
          Whole Mouse Genome G4122A A_52_P404533 X14686.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P404533 X14687.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P404533 AK010663.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P404533 AK168927.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P404533 BC018257.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138341 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104893 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04316885 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04316882 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04316888 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24118365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24118384 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24118341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24118336 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24118380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04958895 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24118383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24118361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04394443 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24118374 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24118367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24118371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24118388 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04962649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24118343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24118389 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04342692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24118338 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04288743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24118386 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24118328 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24118395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04288742 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04400800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04818558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24118368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24118351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04729409 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24118333 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04422056 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24118355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04316888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24118384 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24118339 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24118352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24118394 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04342690 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04400795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24118354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04921639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24118390 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04316882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24118346 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24118349 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10526417 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526415 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408929 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526411 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408930 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404873 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408931 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526416 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526412 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526413 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526414 - -
          Affymetrix HT MG-430 PM Array Plate 1425964_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1422943_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10408928 - -
          Affymetrix Mouse Gene 1.1 ST Array 10526410 - -
          Affymetrix Mouse Gene 1.1 ST Array 10404872 - -
          Mouse Genome 430 2.0 Array 1427854_x_at - -
          Mouse Genome 430 2.0 Array 1425964_x_at - -
          Mouse Genome 430 2.0 Array 1453914_at - -
          Mouse Genome 430 2.0 Array 1427853_a_at - -
          Mouse Genome 430 2.0 Array 1422943_PM_a_at - -
          Mouse Genome 430 2.0 Array 1425964_PM_x_at - -
          Mouse Genome 430 2.0 Array 1427853_PM_a_at - -
          Mouse Genome 430 2.0 Array 1427854_PM_x_at - -
          Mouse Genome 430 2.0 Array 1453914_PM_at - -
          Mouse Genome 430 2.0 Array 1422943_a_at - -
          Affymetrix Mouse Clariom S Array PSR0500014350.mm.1 TC0500001590.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500014350.mm.1 NM_013560.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014350.mm.1 NM_013560.2 Refseq
          Mouse Expression Set 430 1422943_PM_a_at - -
          Mouse Expression Set 430 1425964_x_at - -
          Mouse Expression Set 430 1427853_a_at - -
          Mouse Expression Set 430 1427854_x_at - -
          Mouse Expression Set 430 1427853_PM_a_at - -
          Mouse Expression Set 430 1422943_a_at - -
          Mouse Expression Set 430 1425964_PM_x_at - -
          Mouse Expression Set 430 1427854_PM_x_at - -
          Murine Genome U74v2 Set 92853_PM_at - -
          Murine Genome U74v2 Set 92853_at - -
          Murine Genome U74v2 Set 171538_PM_f_at - -
          Murine Genome U74v2 Set 171538_f_at - -
          Mouse Genome 430A 2.0 Array 1425964_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1425964_x_at - -
          Mouse Genome 430A 2.0 Array 1427854_x_at - -
          Mouse Genome 430A 2.0 Array 1422943_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1422943_a_at - -
          Mouse Genome 430A 2.0 Array 1427853_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1427854_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1427853_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1425964_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1427854_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1422943_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1427854_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1422943_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1427853_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1425964_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1427853_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 485972 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5420300 - -
          MouseWG-6 v2 Expression BeadChip 100580593 - -
          Illumina mouse-6 v1.0 expression beadchip 5420300 - -
          Illumina mouse-6 v1.0 expression beadchip 100580593 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005420300 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2706269 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2706269 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2515964 - -
          mouse-6 v1.1 (Illumina) scl12257.1.1_274-S - -
          mouse-6 v1.1 (Illumina) scl0015507.1_320-S - -
          Illumina Sentrix mouse-6.v1 bead array 100580593 - -
          Illumina Sentrix mouse-6.v1 bead array 5420300 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0015507.1_320-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5420300 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100580593 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2706269 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2706269 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2515964 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0015507.1_320-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0