| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26912795 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26912794 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26912793 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2117689 |
CK377643.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2117689 |
BC027299.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2117689 |
AK218009.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2117689 |
AK163523.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2117689 |
AK212813.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
80546 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
299393 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
777782 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
809901 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
530114 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
214804 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
572887 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
907584 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
356168 |
AK028153.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
356168 |
AK029168.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
356168 |
AK033601.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
356168 |
BC048400.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
356168 |
NM_172966.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
556382 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
48333 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
692743 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
776070 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
122161 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
911084 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
165136 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
4912 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
157806 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
111943 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P102703 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P158939 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2117689 |
CK377643.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2117689 |
BC027299.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2117689 |
AK218009.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2117689 |
AK163523.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2117689 |
AK212813.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26912819 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26912797 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26912800 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26912796 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26912825 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26912802 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26912795 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26912799 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26912808 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26912817 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26912811 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26912807 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26912813 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26912805 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26912816 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P158939 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06964398 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P102703 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P02056821 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P147562 |
AK075603.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P399646 |
BC019572.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P399646 |
BC027299.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P399646 |
BC036150.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P399646 |
AK163523.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P399646 |
AK205767.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P104274 |
AK033298.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P136656 |
- |
- |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P101390 |
- |
- |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P122689 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P158939 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P102703 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08634946 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08420978 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08437186 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08952294 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08634948 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08437184 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26912794 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26912795 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26912793 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912826 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912942 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912914 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912991 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26913004 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912905 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912791 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912868 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912935 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912839 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912849 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912829 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912962 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912858 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26913007 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912915 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912931 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912984 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912901 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912843 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912828 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912807 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912953 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912899 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912857 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912891 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912960 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26913003 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912998 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912792 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912919 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912871 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912834 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912900 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912988 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912982 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912884 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912827 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912985 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912873 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912817 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912944 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912847 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26913009 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912869 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912805 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912896 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912923 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912968 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912851 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912987 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912980 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912799 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912841 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912889 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912970 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912877 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912955 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912940 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912846 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912949 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912803 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912879 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912800 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26913006 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912974 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912844 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26913000 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912948 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912979 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912811 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912836 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912908 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912888 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912831 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912941 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912933 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912882 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912925 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912911 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26913013 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912887 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912813 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912992 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912952 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912808 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912971 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912909 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26913014 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912951 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912999 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26912796 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08714856 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26912860 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08507070 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26912828 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26912845 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08503933 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26912813 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26912803 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08507081 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26912846 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26912811 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08470269 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26912841 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08634948 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26912861 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26912816 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26912843 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26912829 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26912836 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08952294 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26912839 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08478156 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08503937 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08503931 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08507076 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26912799 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26912808 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26912847 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26912851 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08507077 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08420981 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26912834 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26912798 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26912800 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08880934 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08376719 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26912807 |
- |
- |