Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00835117_g1
          See other TMA16 GE Assays ›
          Gene Symbol:
          Tma16
          Gene Name:
          translation machinery associated 16 homolog (S. cerevisiae)
          Gene Aliases:
          1810029B16Rik
          Chromosome Location:
          Chr.8: 66473118 - 66486548 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00835117_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tma16
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025465.2 NP_079741.1 6 - 7 520 95
          GenBank mRNA AK007637.1 - 6 - 7 520 95
          AK009167.1 - 6 - 7 535 95
          AK011532.1 - 6 - 7 531 95
          BC016246.1 - 6 - 7 566 4511374 95
          BC034844.1 - 6 - 7 516 1496036 95

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tma16

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25933302 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25933301 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25933300 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25933298 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982628 BC016246.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982628 BC034844.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982628 NM_025465.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982628 AK007637.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982628 AK009167.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994654 BC016246.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994654 NM_025465.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994654 AK007637.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 861565 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 935445 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 858207 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 654242 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 27271 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 951933 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 134892 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166329 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179303 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139006 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982628 BC016246.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982628 BC034844.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982628 NM_025465.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982628 AK007637.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982628 AK009167.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994654 BC016246.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994654 NM_025465.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994654 AK007637.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25933298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25933291 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25933305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25933307 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25933280 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25933303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25933289 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25933302 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25933285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25933293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25933282 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25933300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25933311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25933287 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25933309 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25933288 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25933295 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25933283 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166329 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139006 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06540348 - -
          Whole Mouse Genome G4122A A_52_P255618 AK051835.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P264525 BC016246.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P306839 AK007637.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139006 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166329 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07154217 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07154228 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25933298 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25933300 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25933301 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25933302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25933285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06855081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06861240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933288 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933307 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06997681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07115048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07240236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933273 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933319 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06855087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06855082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06855090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07408287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25933320 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10578968 - -
          Affymetrix Mouse Gene 1.0 ST Array 10578966 - -
          Affymetrix Mouse Gene 1.0 ST Array 10578965 - -
          Affymetrix Mouse Gene 1.0 ST Array 10578970 - -
          Affymetrix Mouse Gene 1.0 ST Array 10578972 - -
          Affymetrix Mouse Gene 1.0 ST Array 10578971 - -
          Affymetrix Mouse Gene 1.0 ST Array 10578969 - -
          Affymetrix Mouse Gene 1.0 ST Array 10578967 - -
          Affymetrix HT MG-430 PM Array Plate 1431117_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1441400_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1423289_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10578964 - -
          Mouse Genome 430 2.0 Array 1423289_PM_a_at - -
          Mouse Genome 430 2.0 Array 1441400_at - -
          Mouse Genome 430 2.0 Array 1431117_PM_x_at - -
          Mouse Genome 430 2.0 Array 1431117_x_at - -
          Mouse Genome 430 2.0 Array 1441400_PM_at - -
          Mouse Genome 430 2.0 Array 1423289_a_at - -
          Affymetrix Mouse Clariom S Array PSR0800017549.mm.1 NM_025465.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017549.mm.1 NM_025465.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017549.mm.1 TC0800002330.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800017551.mm.1 NM_025465.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017551.mm.1 NM_025465.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017551.mm.1 TC0800002330.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800017554.mm.1 NM_025465.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017554.mm.1 NM_025465.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017554.mm.1 TC0800002330.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800017543.mm.1 TC0800002330.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800017546.mm.1 NM_025465.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017546.mm.1 NM_025465.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017546.mm.1 TC0800002330.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800017545.mm.1 NM_025465.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017545.mm.1 NM_025465.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017545.mm.1 TC0800002330.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800017552.mm.1 TC0800002330.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800017555.mm.1 NM_025465.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017555.mm.1 NM_025465.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017555.mm.1 TC0800002330.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800017548.mm.1 NM_025465.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017548.mm.1 NM_025465.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017548.mm.1 TC0800002330.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800017544.mm.1 NM_025465.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017544.mm.1 NM_025465.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017544.mm.1 TC0800002330.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800017547.mm.1 NM_025465.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017547.mm.1 NM_025465.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017547.mm.1 TC0800002330.mm.1 MTA TC
          Mouse Expression Set 430 1431117_x_at - -
          Mouse Expression Set 430 1423289_a_at - -
          Mouse Expression Set 430 1423289_PM_a_at - -
          Mouse Expression Set 430 1431117_PM_x_at - -
          Mouse Expression Set 430 1441400_at - -
          Mouse Expression Set 430 1441400_PM_at - -
          Mouse Genome 430A 2.0 Array 1431117_x_at - -
          Mouse Genome 430A 2.0 Array 1423289_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1431117_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1423289_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1431117_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1423289_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1423289_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1431117_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1441400_at - -
          Affymetrix HT Mouse Genome 430B Array 1441400_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 341560 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6100451 - -
          MouseWG-6 v2 Expression BeadChip 3870242 - -
          Illumina mouse-6 v1.0 expression beadchip 6100451 - -
          Illumina mouse-6 v1.0 expression beadchip 101990433 - -
          Illumina mouse-6 v1.0 expression beadchip 3870242 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003870242 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006100451 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2647318 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2701207 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1257373 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2701207 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2647318 - -
          mouse-6 v1.1 (Illumina) scl066282.1_268-S - -
          mouse-6 v1.1 (Illumina) scl0066282.1_322-S - -
          Illumina Sentrix mouse-6.v1 bead array 3870242 - -
          Illumina Sentrix mouse-6.v1 bead array 101990433 - -
          Illumina Sentrix mouse-6.v1 bead array 6100451 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0066282.1_322-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl066282.1_268-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3870242 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6100451 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101990433 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2701207 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2647318 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2573998 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2701207 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2647318 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline