Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00835449_g1
          See other IFI27 GE Assays ›
          Gene Symbol:
          Ifi27
          Gene Name:
          interferon, alpha-inducible protein 27
          Gene Aliases:
          1110013J02Rik, 2900026P10Rik, D12Ertd647e, ISG12a, Ifi27l1
          Chromosome Location:
          Chr.12: 103434189 - 103440245 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00835449_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ifi27
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026790.2 NP_081066.2 6 - 7 760 103
          NM_194066.2 NP_919042.2 5 - 6 730 103
          NM_194067.2 NP_919043.1 5 - 6 625 103
          NM_194068.2 NP_919044.1 4 - 5 595 103
          NM_194069.2 NP_919045.1 3 - 4 475 103
          GenBank mRNA AJ566111.1 - 6 - 7 738 103
          AJ566112.1 - 5 - 6 708 103
          AJ566113.1 - 5 - 6 603 103
          AJ566114.1 - 4 - 5 573 103
          AJ566115.1 - 3 - 4 453 103
          AK003665.1 - 5 - 6 564 103
          AK075797.2 - 6 - 7 699 103
          AK146231.1 - 5 - 6 602 103
          AK169926.1 - 6 - 7 690 103
          AK200168.1 - 1 - 2 112 103
          BC125301.1 - 3 - 4 407 40129816 103
          BC125636.1 - 3 - 4 407 40130151 103
          BC128276.1 - 6 - 7 659 40091489 103
          BY443016.1 - 1 - 2 132 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200001030.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ifi27

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          regulation of protein export from nucleus
          RNA polymerase II activating transcription factor binding
          lamin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P86693 AK169926.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P86693 BY443016.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P86693 AJ566113.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P86693 AJ566114.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P86693 AJ566115.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P86693 AK169926.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P86693 BY443016.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P86693 AJ566113.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P86693 AJ566114.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P86693 AJ566115.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 849250 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 700464 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 512217 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 950095 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 91691 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158807 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P86693 AK169926.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P86693 BY443016.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P86693 AJ566113.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P86693 AJ566114.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P86693 AJ566115.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P86693 AK169926.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P86693 BY443016.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P86693 AJ566113.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P86693 AJ566114.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P86693 AJ566115.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28697576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28697573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28697582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28697571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28697578 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28697569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28697580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28697579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28697583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28697584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28697567 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02594107 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158807 - -
          Whole Mouse Genome G4122A A_52_P86693 AK169926.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P86693 BY443016.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P86693 AJ566113.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P86693 AJ566114.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P86693 AJ566115.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P86693 AK169926.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P86693 BY443016.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P86693 AJ566113.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P86693 AJ566114.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P86693 AJ566115.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121789 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158807 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09925167 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09925165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09925159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10174928 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09834611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09992771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09925162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09992785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09925158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09925152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09834613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09834620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09808162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09992791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697583 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10397979 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397977 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397982 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397981 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397980 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397978 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397976 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397983 - -
          Affymetrix HT MG-430 PM Array Plate 1454757_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1452956_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10397975 - -
          Mouse Genome 430 2.0 Array 1454757_PM_s_at - -
          Mouse Genome 430 2.0 Array 1454757_s_at - -
          Mouse Genome 430 2.0 Array 1452956_a_at - -
          Mouse Genome 430 2.0 Array 1452956_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 52668_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 52668_at - -
          Affymetrix Mouse Clariom S Array PSR1200007331.mm.1 NM_026790.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007331.mm.1 NM_026790.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007331.mm.1 NM_194066.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007331.mm.1 NM_194066.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007331.mm.1 NM_194067.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007331.mm.1 NM_194067.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007331.mm.1 NM_194068.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007331.mm.1 NM_194068.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007331.mm.1 NM_194069.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007331.mm.1 NM_194069.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007331.mm.1 TC1200001030.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200007334.mm.1 NM_026790.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007334.mm.1 NM_026790.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007334.mm.1 NM_194066.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007334.mm.1 NM_194066.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007334.mm.1 NM_194067.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007334.mm.1 NM_194067.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007334.mm.1 NM_194068.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007334.mm.1 NM_194068.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007334.mm.1 NM_194069.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007334.mm.1 NM_194069.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007334.mm.1 TC1200001030.mm.1 MTA TC
          Mouse Expression Set 430 1454757_PM_s_at - -
          Mouse Expression Set 430 1454757_s_at - -
          Mouse Expression Set 430 1452956_PM_a_at - -
          Mouse Expression Set 430 1452956_a_at - -
          Murine Genome U74v2 Set 93775_PM_at - -
          Murine Genome U74v2 Set 93775_at - -
          Affymetrix HT Mouse Genome 430B Array 1452956_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1452956_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1454757_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1454757_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 442062 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5290364 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005290364 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2681232 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2681232 - -
          mouse-6 v1.1 (Illumina) scl42842.7.1_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 5290364 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl42842.7.1_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5290364 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2681232 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3125606 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3125606 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2681232 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl42842.7.1_0-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline