Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00835620_m1
          See other SPEER4B GE Assays ›
          Gene Symbol:
          Speer4b
          Gene Name:
          spermatogenesis associated glutamate (E)-rich protein 4b
          Gene Aliases:
          1700081O22Rik, SPEER-4B
          Chromosome Location:
          Chr.5: 27495809 - 27501392 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00835620_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Speer4b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028561.2 NP_082837.1 3 - 4 464 144
          GenBank mRNA AK006975.1 - 3 - 4 513 144
          BC119075.1 - 3 - 4 464 8733824 144

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002130.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Speer4b

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2075067 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P100496 NM_028561.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P100496 AK006975.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P100496 NM_028561.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P100496 AK006975.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2075067 - -
          Whole Mouse Genome G4122A A_52_P610872 NM_028561.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P610872 AK006975.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P23557753 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10528825 - -
          Affymetrix Mouse Gene 1.0 ST Array 10528797 - -
          Affymetrix Mouse Gene 1.0 ST Array 10528814 - -
          Affymetrix Mouse Gene 1.0 ST Array 10519806 - -
          Affymetrix Mouse Gene 1.0 ST Array 10528796 - -
          Affymetrix Mouse Gene 1.0 ST Array 10528819 - -
          Affymetrix Mouse Gene 1.0 ST Array 10528806 - -
          Affymetrix Mouse Gene 1.0 ST Array 10528798 - -
          Affymetrix Mouse Gene 1.0 ST Array 10528809 - -
          Affymetrix Mouse Gene 1.0 ST Array 10528805 - -
          Affymetrix Mouse Gene 1.0 ST Array 10528839 - -
          Affymetrix Mouse Gene 1.0 ST Array 10528808 - -
          Affymetrix Mouse Gene 1.0 ST Array 10528816 - -
          Affymetrix Mouse Gene 1.0 ST Array 10528835 - -
          Affymetrix Mouse Gene 1.0 ST Array 10528838 - -
          Affymetrix Mouse Gene 1.0 ST Array 10519810 - -
          Affymetrix Mouse Gene 1.0 ST Array 10528836 - -
          Affymetrix Mouse Gene 1.0 ST Array 10528823 - -
          Affymetrix Mouse Gene 1.0 ST Array 10528837 - -
          Affymetrix Mouse Gene 1.0 ST Array 10528817 - -
          Affymetrix Mouse Gene 1.0 ST Array 10528826 - -
          Affymetrix Mouse Gene 1.0 ST Array 10528824 - -
          Affymetrix Mouse Gene 1.0 ST Array 10528822 - -
          Affymetrix Mouse Gene 1.0 ST Array 10528812 - -
          Affymetrix Mouse Gene 1.0 ST Array 10528795 - -
          Affymetrix Mouse Gene 1.0 ST Array 10528811 - -
          Affymetrix HT MG-430 PM Array Plate 1429992_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10528794 - -
          Affymetrix Mouse Gene 1.1 ST Array 10528804 - -
          Affymetrix Mouse Gene 1.1 ST Array 10528810 - -
          Affymetrix Mouse Gene 1.1 ST Array 10528815 - -
          Affymetrix Mouse Gene 1.1 ST Array 10528821 - -
          Affymetrix Mouse Gene 1.1 ST Array 10528834 - -
          Mouse Genome 430 2.0 Array 1429993_s_at - -
          Mouse Genome 430 2.0 Array 1429992_at - -
          Mouse Genome 430 2.0 Array 1429993_PM_s_at - -
          Mouse Genome 430 2.0 Array 1429992_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500019359.mm.1 TC0500002130.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019359.mm.1 NM_028561.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019359.mm.1 NM_028561.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019363.mm.1 TC0500002130.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019363.mm.1 NM_028561.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019363.mm.1 NM_028561.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019358.mm.1 TC0500002130.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019358.mm.1 NM_028561.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019358.mm.1 NM_028561.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019361.mm.1 TC0500002130.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019361.mm.1 NM_028561.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019361.mm.1 NM_028561.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019362.mm.1 TC0500002130.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019362.mm.1 NM_028561.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019362.mm.1 NM_028561.2 Refseq
          Mouse Expression Set 430 1429992_PM_at - -
          Mouse Expression Set 430 1429993_PM_s_at - -
          Mouse Expression Set 430 1429993_s_at - -
          Mouse Expression Set 430 1429992_at - -
          Affymetrix HT Mouse Genome 430B Array 1429992_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429992_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 339807 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 106040692 - -
          Illumina mouse-6 v1.0 expression beadchip 2120673 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002120673 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2697074 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2697074 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232531 - -
          mouse-6 v1.1 (Illumina) scl0073526.1_245-S - -
          mouse-6 v1.1 (Illumina) GI_21735468-S - -
          Illumina Sentrix mouse-6.v1 bead array 2120673 - -
          Illumina Sentrix mouse-6.v1 bead array 106040692 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0073526.1_245-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106040692 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2120673 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2542607 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2542607 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline