Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00835933_s1
          See other VMN1R25 GE Assays ›
          Gene Symbol:
          Vmn1r25
          Gene Name:
          vomeronasal 1 receptor 25
          Gene Aliases:
          V1rc8
          Chromosome Location:
          Chr.6: 57978394 - 57979302 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00835933_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn1r25
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053238.2 NP_444468.2 1 773 96

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn1r25

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          sensory perception of chemical stimulus
          receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P110108 NM_053238.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P110108 NM_053238.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P110108 NM_053238.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P110108 NM_053238.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P110108 NM_053238.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P110108 NM_053238.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24480692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24480735 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24480702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24480733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24480732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24480711 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24480720 - -
          Whole Mouse Genome G4122A A_51_P110108 NM_053238.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P110108 NM_053238.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115156 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134167 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05227592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24480751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24480733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24480716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05372259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24480680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05848546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05507184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24480702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05227590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24480711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05146119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24480733 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10545031 - -
          Affymetrix HT MG-430 PM Array Plate 1422388_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10545030 - -
          Mouse Genome 430 2.0 Array 1422388_PM_at - -
          Mouse Genome 430 2.0 Array 1422388_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 113865_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 113865_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600018388.mm.1 TC0600002362.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600018388.mm.1 NM_053238.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018388.mm.1 NM_053238.2 Refseq
          Mouse Expression Set 430 1422395_PM_at - -
          Mouse Expression Set 430 1422395_at - -
          Mouse Expression Set 430 1422388_at - -
          Mouse Expression Set 430 1422388_PM_at - -
          Mouse Genome 430A 2.0 Array 1422388_at - -
          Mouse Genome 430A 2.0 Array 1422395_at - -
          Mouse Genome 430A 2.0 Array 1422388_PM_at - -
          Mouse Genome 430A 2.0 Array 1422395_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422388_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422388_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 491834 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 100070347 - -
          Illumina mouse-6 v1.0 expression beadchip 3940408 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003940408 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2482362 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2482362 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1259626 - -
          mouse-6 v1.1 (Illumina) GI_16716558-S - -
          mouse-6 v1.1 (Illumina) scl0113865.1_49-S - -
          Illumina Sentrix mouse-6.v1 bead array 3940408 - -
          Illumina Sentrix mouse-6.v1 bead array 100070347 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0113865.1_49-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100070347 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3940408 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2482362 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2482362 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline