Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00836196_s1
          See other VMN1R200 GE Assays ›
          Gene Symbol:
          Vmn1r200
          Gene Name:
          vomeronasal 1 receptor 200
          Gene Aliases:
          V1rh3
          Chromosome Location:
          Chr.13: 22395029 - 22395967 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00836196_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn1r200
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134212.1 NP_598973.1 1 263 154
          GenBank mRNA BC131902.1 - 1 345 40130179 154
          BC131904.1 - 1 345 40130181 154

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn1r200

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          response to pheromone
          pheromone binding
          pheromone receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P325006 BC131904.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P325006 NM_134212.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P325006 BC131902.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P325006 BC131904.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P325006 NM_134212.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P325006 BC131902.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P325006 BC131904.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P325006 NM_134212.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P325006 BC131902.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P325006 BC131904.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P325006 NM_134212.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P325006 BC131902.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P325006 BC131904.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P325006 NM_134212.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P325006 BC131902.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P325006 BC131904.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P325006 NM_134212.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P325006 BC131902.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28877603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28877611 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28877566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28877577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28877589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28877574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28877576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28877605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28877580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28877581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28877579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28877606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28877586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28877607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28877575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28877568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28877592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28877587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28877609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28877578 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28877594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28877612 - -
          Whole Mouse Genome G4122A A_51_P325006 BC131904.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P325006 NM_134212.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P325006 BC131902.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P325006 BC131904.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P325006 NM_134212.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P325006 BC131902.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132489 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137165 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139510 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10957764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28877551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11070078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10507590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877614 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877557 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10604252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877642 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10507592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10604259 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877627 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877563 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10604257 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10604263 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28877581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11076830 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10403997 - -
          Affymetrix Mouse Gene 1.1 ST Array 10403996 - -
          Affymetrix Mouse Clariom S Array PSR1300001284.mm.1 NM_134212.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001284.mm.1 NM_134212.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001284.mm.1 TC1300000205.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300001283.mm.1 TC1300000205.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 452998 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 105390377 - -
          Illumina mouse-6 v1.0 expression beadchip 2350088 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002350088 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1231031 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1220756 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1231031 - -
          mouse-6 v1.1 (Illumina) GI_21717736-S - -
          mouse-6 v1.1 (Illumina) scl12349.1.1_100-S - -
          Illumina Sentrix mouse-6.v1 bead array 2350088 - -
          Illumina Sentrix mouse-6.v1 bead array 105390377 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl12349.1.1_100-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105390377 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2350088 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1231031 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2940118 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1231031 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2940118 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline