Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00836333_m1
          See other D10JHU81E GE Assays ›
          Gene Symbol:
          D10Jhu81e
          Gene Name:
          DNA segment, Chr 10, Johns Hopkins University 81 expressed
          Gene Aliases:
          C21orf33, ES1
          Chromosome Location:
          Chr.10: 78162067 - 78169768 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00836333_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: D10Jhu81e
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138601.2 NP_613067.1 5 - 6 557 94
          GenBank mRNA AK003866.1 - 5 - 6 558 94
          BC013475.1 - 5 - 6 550 4237589 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002442.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene D10Jhu81e

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27272487 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27272484 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27272485 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27272488 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27272486 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2164909 AK003866.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2164909 BC013475.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2164909 NM_138601.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1991109 AK003866.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1991109 NM_138601.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 334443 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 608726 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 428656 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 451922 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 342754 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 602406 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P177483 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1991109 AK003866.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1991109 NM_138601.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2164909 AK003866.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2164909 BC013475.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2164909 NM_138601.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27272483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27272468 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27272484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27272474 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27272472 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27272471 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27272489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27272478 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27272492 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27272480 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27272488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27272496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27272494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27272486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27272469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27272490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27272475 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P177483 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07117412 - -
          Whole Mouse Genome G4122A A_52_P498167 AK003866.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P498167 BC013475.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P498167 NM_138601.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P118320 AK003866.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P118320 BC013475.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P118320 NM_138601.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P177483 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08817503 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08817509 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08817497 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08817494 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08817495 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27272484 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27272487 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27272488 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27272485 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27272486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27272395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08817495 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08866252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08680894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08382615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272480 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08382618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272436 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272461 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272472 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08617060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272452 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272454 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08866257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272439 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27272494 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10370415 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370414 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370421 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370416 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370417 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370420 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370418 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370419 - -
          Affymetrix HT MG-430 PM Array Plate 1449000_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10370413 - -
          Mouse Genome 430 2.0 Array 1449000_at - -
          Mouse Genome 430 2.0 Array 1449000_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 28295_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 28295_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000017757.mm.1 NM_138601.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017757.mm.1 NM_138601.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017757.mm.1 TC1000002442.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000017758.mm.1 NM_138601.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017758.mm.1 NM_138601.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017758.mm.1 TC1000002442.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000017759.mm.1 NM_138601.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017759.mm.1 NM_138601.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017759.mm.1 TC1000002442.mm.1 MTA TC
          Mouse Expression Set 430 1449000_at - -
          Mouse Expression Set 430 1449000_PM_at - -
          Murine Genome U74v2 Set 96757_PM_at - -
          Murine Genome U74v2 Set 96757_at - -
          Mouse Genome 430A 2.0 Array 1449000_at - -
          Mouse Genome 430A 2.0 Array 1449000_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 665569 - -
          ABI Mouse Genome Survey Microarray 907102 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6840400 - -
          Illumina mouse-6 v1.0 expression beadchip 6840400 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006840400 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2759521 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2759521 - -
          mouse-6 v1.1 (Illumina) scl37776.6_58-S - -
          Illumina Sentrix mouse-6.v1 bead array 6840400 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37776.6_58-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6840400 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2991545 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2991545 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl37776.6_58-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline