Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00836474_m1
          See other ZFP932 GE Assays ›
          Gene Symbol:
          Zfp932
          Gene Name:
          zinc finger protein 932
          Gene Aliases:
          2310001H12Rik, Znf431
          Chromosome Location:
          Chr.5: 109996527 - 110010411 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00836474_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp932
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145563.2 NP_663538.2 3 - 4 293 141
          GenBank mRNA AK032220.1 - 2 - 3 422 141
          BC012405.1 - 3 - 4 279 141

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001128.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp932

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          cell differentiation
          negative regulation of sequence-specific DNA binding transcription factor activity
          negative regulation of osteoblast differentiation
          core promoter sequence-specific DNA binding
          nucleic acid binding
          chromatin binding
          protein binding
          histone deacetylase binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2032278 AK009062.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2032294 BC012405.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2032294 NM_145563.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2032294 AK032220.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2032294 BC012405.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2032294 NM_145563.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2032294 AK032220.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2032278 AK009062.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05692207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977429 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10524085 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524088 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524080 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524081 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532312 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532307 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524084 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524083 - -
          Affymetrix HT MG-430 PM Array Plate 1424872_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10532305 - -
          Affymetrix Mouse Gene 1.1 ST Array 10532310 - -
          Affymetrix Mouse Gene 1.1 ST Array 10524082 - -
          Affymetrix Mouse Gene 1.1 ST Array 10524079 - -
          Mouse Genome 430 2.0 Array 1424872_PM_at - -
          Mouse Genome 430 2.0 Array 1424872_at - -
          Affymetrix Mouse Clariom S Array PSR0500009605.mm.1 TC0500001128.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500009605.mm.1 NM_145563.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500009595.mm.1 TC0500001128.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500009595.mm.1 NM_145563.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500009603.mm.1 TC0500001128.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500009603.mm.1 NM_145563.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500009601.mm.1 TC0500001128.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500009601.mm.1 NM_145563.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500009600.mm.1 TC0500001128.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500009600.mm.1 NM_145563.2 Refseq
          Murine Genome U74v2 Set 92338_f_at - -
          Murine Genome U74v2 Set 92338_PM_f_at - -
          Murine Genome U74v2 Set 140565_PM_f_at - -
          Murine Genome U74v2 Set 140565_f_at - -
          Affymetrix HT Mouse Genome 430A Array 1424872_at - -
          Affymetrix HT Mouse Genome 430A Array 1424872_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 320321 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5220059 - -
          Illumina mouse-6 v1.0 expression beadchip 104610039 - -
          Illumina mouse-6 v1.0 expression beadchip 5390279 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005220059 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005390279 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2720620 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2687845 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2720620 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232900 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2687845 - -
          mouse-6 v1.1 (Illumina) GI_21704131-S - -
          mouse-6 v1.1 (Illumina) scl0069504.2_283-S - -
          mouse-6 v1.1 (Illumina) scl0069504.1_70-S - -
          Illumina Sentrix mouse-6.v1 bead array 5220059 - -
          Illumina Sentrix mouse-6.v1 bead array 5390279 - -
          Illumina Sentrix mouse-6.v1 bead array 104610039 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0069504.1_70-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0069504.2_283-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5390279 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5220059 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104610039 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2687845 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1232900 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2720620 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0069504.2_283-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0069504.1_70-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline