Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00836694_s1
          See other OLFR129 GE Assays ›
          Gene Symbol:
          Olfr129
          Gene Name:
          olfactory receptor 129
          Gene Aliases:
          MOR263-9
          Chromosome Location:
          Chr.17: 38054599 - 38059873 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00836694_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr129
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146327.2 NP_666439.2 4 970 136

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr129

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006240 NM_146327.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 481254 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134129 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006240 NM_146327.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P200114 NM_146327.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170868 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170947 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170934 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170878 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170910 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170933 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170891 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170901 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170936 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170894 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170950 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170880 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P134129 - -
          Whole Mouse Genome G4122A A_51_P200114 NM_146327.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P134129 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13075048 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13075053 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13075064 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13075083 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13075091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170889 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170862 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170889 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13071359 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13353212 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170856 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13095791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170923 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13383189 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13383190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170920 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170833 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13411183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170895 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170899 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13353213 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10450909 - -
          Affymetrix Mouse Gene 1.1 ST Array 10450908 - -
          Affymetrix Mouse Clariom S Array PSR1700019635.mm.1 NM_146327.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019635.mm.1 NM_146327.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019635.mm.1 TC1700002087.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019636.mm.1 NM_146327.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019636.mm.1 NM_146327.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019636.mm.1 TC1700002087.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019638.mm.1 NM_146327.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019638.mm.1 NM_146327.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019638.mm.1 TC1700002087.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019640.mm.1 NM_146327.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019640.mm.1 NM_146327.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019640.mm.1 TC1700002087.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019639.mm.1 NM_146327.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019639.mm.1 NM_146327.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019639.mm.1 TC1700002087.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 332683 - -
          ABI Mouse Genome Survey Microarray 542645 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2340731 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002340731 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2666043 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2666043 - -
          mouse-6 v1.1 (Illumina) scl7844.1.1_53-S - -
          Illumina Sentrix mouse-6.v1 bead array 2340731 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl7844.1.1_53-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2340731 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2666043 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2666043 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline