Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00836704_s1
          See other OLFR954 GE Assays ›
          Gene Symbol:
          Olfr954
          Gene Name:
          olfactory receptor 954
          Gene Aliases:
          MOR171-42, MOR171-53
          Chromosome Location:
          Chr.9: 39461433 - 39462377 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00836704_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr954
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146331.1 NP_666443.1 1 751 158

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr954

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963570 NM_146331.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963570 NM_146331.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438570 - -
          Whole Mouse Genome G4122A A_51_P319470 NR_033620.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P319470 BC051435.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127039 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07641836 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07641838 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07641860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07505348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08174783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08154945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08174763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438547 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10584493 - -
          Affymetrix Mouse Gene 1.0 ST Array 10584491 - -
          Affymetrix Mouse Gene 1.1 ST Array 10584490 - -
          Affymetrix Mouse Gene 1.1 ST Array 10584492 - -
          Affymetrix Mouse Clariom S Array PSR0900002895.mm.1 NM_146331.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002895.mm.1 NM_146331.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002895.mm.1 TC0900000429.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 754993 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1230021 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001230021 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1231432 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1231432 - -
          mouse-6 v1.1 (Illumina) scl0258328.1_208-S - -
          Illumina Sentrix mouse-6.v1 bead array 1230021 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258328.1_208-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1230021 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2690093 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2690093 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline