Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00836732_s1
          See other OLFR1265 GE Assays ›
          Gene Symbol:
          Olfr1265
          Gene Name:
          olfactory receptor 1265
          Gene Aliases:
          MOR228-2
          Chromosome Location:
          Chr.2: 90036921 - 90037850 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00836732_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1265
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146343.1 NP_666455.1 1 568 161

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1265

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1988989 NM_146343.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980094 NM_146343.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980094 NM_146343.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1988989 NM_146343.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415649 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415647 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415626 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106673 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104090 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123385 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02204280 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02204294 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02204274 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02204292 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02204290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415642 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02080222 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415647 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415618 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02204292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415657 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415653 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415642 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415616 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415646 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415666 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01562395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415650 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415636 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02080214 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01562403 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415665 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484864 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473642 - -
          Affymetrix Mouse Gene 1.1 ST Array 10473641 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484863 - -
          Affymetrix Mouse Clariom S Array PSR0200010165.mm.1 NM_146343.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010165.mm.1 NM_146343.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010165.mm.1 TC0200001343.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 395910 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 870050 - -
          Illumina mouse-6 v1.0 expression beadchip 870050 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000870050 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1257124 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1257124 - -
          mouse-6 v1.1 (Illumina) scl0258340.1_43-S - -
          Illumina Sentrix mouse-6.v1 bead array 870050 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258340.1_43-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 870050 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1257124 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1257124 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0