Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00836895_s1
          See other OLFR469 GE Assays ›
          Gene Symbol:
          Olfr469
          Gene Name:
          olfactory receptor 469
          Gene Aliases:
          MOR204-21
          Chromosome Location:
          Chr.7: 107822523 - 107823467 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00836895_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr469
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146426.1 NP_666637.1 1 736 177

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr469

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of smell
          response to stimulus
          detection of chemical stimulus involved in sensory perception of smell
          signal transducer activity
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2050828 NM_146426.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2050828 NM_146426.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P313007 NM_146426.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25401802 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25401831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25401818 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25401809 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25401819 - -
          Whole Mouse Genome G4122A A_52_P313007 NM_146426.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06000090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25401868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25401881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25401852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25401856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25401818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25401799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25401797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25401809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25401858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25401860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25401884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25401886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25401869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25401883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25401887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25401880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25401862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25401867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25401863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25401841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25401802 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06171801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25401800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25401842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25401818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25401819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25401797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06666563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06644959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25401799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25401831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25401809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25401852 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566663 - -
          Affymetrix Mouse Gene 1.1 ST Array 10566662 - -
          Affymetrix Mouse Clariom S Array PSR0700033520.mm.1 NM_146426.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033520.mm.1 NM_146426.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033520.mm.1 TC0700004040.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2320066 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002320066 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2775879 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2775879 - -
          mouse-6 v1.1 (Illumina) scl0258418.1_214-S - -
          Illumina Sentrix mouse-6.v1 bead array 2320066 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258418.1_214-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2320066 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2805042 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2775879 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2805042 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0