Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00836927_s1
          See other OLFR1314 GE Assays ›
          Gene Symbol:
          Olfr1314
          Gene Name:
          olfactory receptor 1314
          Gene Aliases:
          MOR245-2
          Chromosome Location:
          Chr.2: 112091761 - 112092699 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00836927_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1314
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146450.2 NP_666661.2 1 41 105

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1314

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P629709 NM_146450.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P629709 NM_146450.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174050 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P629709 NM_146450.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P629709 NM_146450.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P629709 NM_146450.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P629709 NM_146450.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174050 - -
          Whole Mouse Genome G4122A A_52_P629709 NM_146450.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P629709 NM_146450.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131840 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01484513 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01484525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527322 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02035006 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527339 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527335 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01484534 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527346 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01484509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02035004 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01484508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527342 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527299 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527298 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527344 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527333 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527297 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01484520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527324 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527310 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10485814 - -
          Affymetrix Mouse Gene 1.1 ST Array 10485813 - -
          Affymetrix Mouse Clariom S Array PSR0200036251.mm.1 NM_146450.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200036251.mm.1 NM_146450.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200036251.mm.1 TC0200004342.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 622835 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6290551 - -
          Illumina mouse-6 v1.0 expression beadchip 6290551 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006290551 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2609919 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2609919 - -
          mouse-6 v1.1 (Illumina) scl18870.1.118_4-S - -
          Illumina Sentrix mouse-6.v1 bead array 6290551 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl18870.1.118_4-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6290551 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2609919 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2609919 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline