Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00836974_s1
          See other OLFR1392 GE Assays ›
          Gene Symbol:
          Olfr1392
          Gene Name:
          olfactory receptor 1392
          Gene Aliases:
          MOR256-25
          Chromosome Location:
          Chr.11: 49293297 - 49294322 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00836974_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1392
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146470.2 NP_666681.1 1 175 224
          GenBank mRNA BC120614.1 - 1 171 40129537 224
          BC120616.1 - 1 175 40129539 224

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1392

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27782378 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27782415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27782414 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27782370 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27782385 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27782399 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27782420 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27782366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27782371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27782391 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27782382 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27782398 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27782393 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100651 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114053 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09095242 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09095239 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09095238 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09095231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27782390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09095245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09095230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782422 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09107661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09107662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09093821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782381 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09614405 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09457452 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782385 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09095229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09107660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09107672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782377 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782334 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08993941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27782362 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10375566 - -
          Affymetrix Mouse Gene 1.0 ST Array 10375546 - -
          Affymetrix Mouse Gene 1.0 ST Array 10375564 - -
          Affymetrix Mouse Gene 1.1 ST Array 10375565 - -
          Affymetrix Mouse Gene 1.1 ST Array 10375563 - -
          Affymetrix Mouse Gene 1.1 ST Array 10375545 - -
          Affymetrix Mouse Clariom S Array PSR1100003886.mm.1 NM_146470.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100003886.mm.1 NM_146470.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100003886.mm.1 TC1100000503.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 590571 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6590025 - -
          Illumina mouse-6 v1.0 expression beadchip 6590025 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006590025 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2705623 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2701018 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2705623 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2701018 - -
          mouse-6 v1.1 (Illumina) scl0258462.1_309-S - -
          Illumina Sentrix mouse-6.v1 bead array 6590025 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258462.1_309-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6590025 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2705623 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline