Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00837023_s1
          See other OLFR492 GE Assays ›
          Gene Symbol:
          Olfr492
          Gene Name:
          olfactory receptor 492
          Gene Aliases:
          MOR204-18
          Chromosome Location:
          Chr.7: 108322730 - 108323674 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00837023_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr492
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146497.1 NP_666708.1 1 269 112

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr492

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of smell
          response to stimulus
          detection of chemical stimulus involved in sensory perception of smell
          signal transducer activity
          G-protein coupled receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P237957 NM_146497.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P237957 NM_146497.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 28380 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118238 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P237957 NM_146497.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P237957 NM_146497.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P237957 NM_146497.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P237957 NM_146497.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403040 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P118238 - -
          Whole Mouse Genome G4122A A_52_P237957 NM_146497.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P237957 NM_146497.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101599 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P118238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06186976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403036 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566689 - -
          Affymetrix Mouse Gene 1.1 ST Array 10566688 - -
          Affymetrix Mouse Clariom S Array PSR0700033540.mm.1 NM_146497.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033540.mm.1 NM_146497.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033540.mm.1 TC0700004054.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 777056 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6130300 - -
          Illumina mouse-6 v1.0 expression beadchip 6130300 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006130300 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2679373 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2679373 - -
          mouse-6 v1.1 (Illumina) scl0258490.1_8-S - -
          Illumina Sentrix mouse-6.v1 bead array 6130300 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258490.1_8-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6130300 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2679373 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2679373 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0