Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00837118_s1
          See other OLFR536 GE Assays ›
          Gene Symbol:
          Olfr536
          Gene Name:
          olfactory receptor 536
          Gene Aliases:
          MOR252-1, MOR252-4
          Chromosome Location:
          Chr.7: 140500853 - 140507318 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00837118_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr536
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146520.2 NP_666731.1 4 290 196
          GenBank mRNA BC051116.1 - 4 290 6307473 196

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr536

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980771 BC051116.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980771 NM_146520.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P557059 BC051116.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P557059 NM_146520.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P557059 BC051116.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P557059 NM_146520.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 295422 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 798229 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 723087 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 189103 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 86183 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119901 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980771 BC051116.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980771 NM_146520.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P557059 BC051116.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P557059 NM_146520.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P557059 BC051116.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P557059 NM_146520.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P557059 BC051116.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P557059 NM_146520.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P557059 BC051116.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P557059 NM_146520.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25583975 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25583987 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25583974 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25583977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25583994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25583999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25583998 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25584000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25583982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25583973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25583979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25583981 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25583983 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25583976 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25583997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25583995 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P119901 - -
          Whole Mouse Genome G4122A A_52_P557059 BC051116.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P557059 NM_146520.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P557059 BC051116.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P557059 NM_146520.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P119901 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06656204 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06656208 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06656210 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06656209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25584000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25583948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06545441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06656203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06559769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06529384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05927966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25584000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05927962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25583997 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10568965 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568966 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568967 - -
          Affymetrix Mouse Gene 1.0 ST Array 10558646 - -
          Affymetrix Mouse Gene 1.1 ST Array 10558645 - -
          Affymetrix Mouse Gene 1.1 ST Array 10568964 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 884438 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6660286 - -
          MouseWG-6 v2 Expression BeadChip 1780400 - -
          MouseWG-6 v2 Expression BeadChip 4730619 - -
          Illumina mouse-6 v1.0 expression beadchip 1780400 - -
          Illumina mouse-6 v1.0 expression beadchip 6660286 - -
          Illumina mouse-6 v1.0 expression beadchip 4730619 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006660286 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004730619 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001780400 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2674023 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2600483 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1231381 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2674023 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2600483 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1231381 - -
          mouse-6 v1.1 (Illumina) scl0258513.1_147-S - -
          mouse-6 v1.1 (Illumina) scl00258513.1_42-S - -
          mouse-6 v1.1 (Illumina) scl00258513.2_43-S - -
          Illumina Sentrix mouse-6.v1 bead array 1780400 - -
          Illumina Sentrix mouse-6.v1 bead array 6660286 - -
          Illumina Sentrix mouse-6.v1 bead array 4730619 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00258513.1_42-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00258513.2_43-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258513.1_147-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1780400 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6660286 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4730619 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2600483 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2674023 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1231381 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2600483 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1231381 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2674023 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline