Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00837168_s1
          See other OLFR1170 GE Assays ›
          Gene Symbol:
          Olfr1170
          Gene Name:
          olfactory receptor 1170
          Gene Aliases:
          MOR174-12
          Chromosome Location:
          Chr.2: 88224080 - 88225030 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00837168_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1170
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146532.1 NP_666743.1 1 635 218
          GenBank mRNA BC127994.1 - 1 639 40135743 218

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1170

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P417727 NM_146532.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P417727 BC127994.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P417727 NM_146532.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P417727 BC127994.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P417727 NM_146532.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P417727 BC127994.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P417727 NM_146532.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P417727 BC127994.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P417727 NM_146532.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P417727 BC127994.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P417727 NM_146532.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P417727 BC127994.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P417727 NM_146532.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P417727 BC127994.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P417727 NM_146532.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P417727 BC127994.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21408942 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408942 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408930 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408927 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408903 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408925 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408960 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02022905 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01913174 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02022915 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408936 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408928 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408901 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408950 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01913177 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408898 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408956 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408904 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408937 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408906 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408893 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21408957 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484727 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484726 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 903582 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1580156 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001580156 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2633839 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2633839 - -
          mouse-6 v1.1 (Illumina) scl00258525.1_37-S - -
          Illumina Sentrix mouse-6.v1 bead array 1580156 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00258525.1_37-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1580156 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2633839 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2633839 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline