Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00837202_s1
          See other OLFR775 GE Assays ›
          Gene Symbol:
          Olfr775
          Gene Name:
          olfactory receptor 775
          Gene Aliases:
          MOR111-6, MOR111-7, Olfr1518
          Chromosome Location:
          Chr.10: 129250536 - 129251474 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00837202_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr775
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146545.2 NP_666756.2 1 551 100

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P138646 NM_146545.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P138646 NM_146545.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P642360 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545189 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545196 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545176 - -
          Whole Mouse Genome G4122A A_52_P642360 - -
          Whole Mouse Genome G4122A A_51_P185275 NM_001011822.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129925 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138646 NM_146545.2 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08939939 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08939940 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08939949 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08543654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08209022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08543667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08543655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545183 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10367480 - -
          Affymetrix Mouse Gene 1.1 ST Array 10367479 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 751610 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1340463 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001340463 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2643751 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2643751 - -
          mouse-6 v1.1 (Illumina) scl00258538.1_10-S - -
          Illumina Sentrix mouse-6.v1 bead array 1340463 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00258538.1_10-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1340463 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2643751 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2643751 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline