Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00837220_s1
          See other OLFR800 GE Assays ›
          Gene Symbol:
          Olfr800
          Gene Name:
          olfactory receptor 800
          Gene Aliases:
          MOR114-7
          Chromosome Location:
          Chr.10: 129659808 - 129660743 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00837220_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr800
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146548.1 NP_666759.1 1 627 197
          GenBank mRNA BC134373.1 - 1 631 40134856 197

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000001649.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr800

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2014675 BC134373.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2014675 NM_146548.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2014675 BC134373.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2014675 NM_146548.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P115896 BC134373.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P115896 NM_146548.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P115895 BC134373.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P115895 NM_146548.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546682 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546685 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546669 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546683 - -
          Whole Mouse Genome G4122A A_51_P115896 BC134373.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115896 NM_146548.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115895 BC134373.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115895 NM_146548.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08465297 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08465287 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08465301 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08465276 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08465317 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08465299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08566212 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546659 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10367514 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373625 - -
          Affymetrix Mouse Gene 1.1 ST Array 10373624 - -
          Affymetrix Mouse Gene 1.1 ST Array 10367513 - -
          Affymetrix Mouse Clariom S Array PSR1000012047.mm.1 NM_146548.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000012047.mm.1 NM_146548.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000012047.mm.1 TC1000001649.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 679116 - -
          ABI Mouse Genome Survey Microarray 753772 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1170040 - -
          MouseWG-6 v2 Expression BeadChip 107100685 - -
          Illumina mouse-6 v1.0 expression beadchip 1170040 - -
          Illumina mouse-6 v1.0 expression beadchip 107100685 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001170040 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2613796 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1252405 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2613796 - -
          mouse-6 v1.1 (Illumina) GI_22203788-S - -
          mouse-6 v1.1 (Illumina) scl0258541.1_56-S - -
          Illumina Sentrix mouse-6.v1 bead array 1170040 - -
          Illumina Sentrix mouse-6.v1 bead array 107100685 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258541.1_56-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1170040 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 107100685 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2613796 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2613796 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline