Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00837226_s1
          See other OLFR810 GE Assays ›
          Gene Symbol:
          Olfr810
          Gene Name:
          olfactory receptor 810
          Gene Aliases:
          MOR113-4
          Chromosome Location:
          Chr.10: 129790649 - 129791587 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00837226_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr810
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146550.1 NP_666761.1 1 263 198
          GenBank mRNA BC120749.1 - 1 290 40129672 198
          BC120751.1 - 1 290 40129674 198

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr810

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P490071 BC120749.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P490071 NM_146550.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P490071 BC120751.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P490071 BC120749.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P490071 NM_146550.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P490071 BC120751.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P490071 BC120749.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P490071 NM_146550.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P490071 BC120751.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P490071 BC120749.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P490071 NM_146550.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P490071 BC120751.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P490071 BC120749.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P490071 NM_146550.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P490071 BC120751.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P490071 BC120749.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P490071 NM_146550.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P490071 BC120751.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547318 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547364 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547321 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547304 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547314 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547352 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547358 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547360 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547315 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547277 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547282 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547309 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547279 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547310 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547295 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547292 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547312 - -
          Whole Mouse Genome G4122A A_52_P490071 BC120749.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P490071 NM_146550.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P490071 BC120751.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P490071 BC120749.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P490071 NM_146550.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P490071 BC120751.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133245 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08926372 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08717723 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08369909 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08969787 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08926370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547363 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547314 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08272476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547358 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08252816 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547273 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08369910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547338 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08868280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547304 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10373643 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373644 - -
          Affymetrix Mouse Gene 1.1 ST Array 10373642 - -
          Affymetrix Mouse Clariom S Array PSR1000023963.mm.1 TC1000003191.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000023964.mm.1 NM_146550.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023964.mm.1 NM_146550.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023964.mm.1 TC1000003191.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000023965.mm.1 TC1000003191.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 874219 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1090594 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001090594 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2637389 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2637389 - -
          mouse-6 v1.1 (Illumina) scl0258543.1_5-S - -
          Illumina Sentrix mouse-6.v1 bead array 1090594 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258543.1_5-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1090594 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2637389 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2637389 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline