Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00837259_sH
          See other OLFR805 GE Assays ›
          Gene Symbol:
          Olfr805
          Gene Name:
          olfactory receptor 805
          Gene Aliases:
          MOR110-4
          Chromosome Location:
          Chr.10: 129722583 - 129723542 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00837259_sH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr805
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146555.1 NP_666766.1 1 549 174
          GenBank mRNA BC111816.1 - 1 548 174

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr805

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P533040 BC111816.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P533040 NM_146555.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P533040 BC111816.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P533040 NM_146555.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140503 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P533040 BC111816.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P533040 NM_146555.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P533040 BC111816.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P533040 NM_146555.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P533040 BC111816.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P533040 NM_146555.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P533040 BC111816.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P533040 NM_146555.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546975 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546996 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546970 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546987 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546978 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546991 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546974 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546984 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546981 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546971 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546985 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27547004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546959 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546989 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546972 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140503 - -
          Whole Mouse Genome G4122A A_52_P533040 BC111816.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P533040 NM_146555.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P533040 BC111816.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P533040 NM_146555.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123130 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109222 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140503 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08227480 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08834710 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08834720 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08834729 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08834719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27547000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08850147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08674449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08674448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08888318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27547005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546999 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10373637 - -
          Affymetrix Mouse Gene 1.1 ST Array 10373636 - -
          Affymetrix Mouse Clariom S Array PSR1000023958.mm.1 NM_146555.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023958.mm.1 NM_146555.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023958.mm.1 TC1000003188.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 895768 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2760739 - -
          Illumina mouse-6 v1.0 expression beadchip 2760739 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002760739 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2736901 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2736901 - -
          mouse-6 v1.1 (Illumina) scl5158.1.1_9-S - -
          Illumina Sentrix mouse-6.v1 bead array 2760739 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl5158.1.1_9-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2760739 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2736901 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline