Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00837298_s1
          See other OLFR698 GE Assays ›
          Gene Symbol:
          Olfr698
          Gene Name:
          olfactory receptor 698
          Gene Aliases:
          MOR283-3
          Chromosome Location:
          Chr.7: 106752451 - 106753386 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00837298_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr698
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146602.2 NP_666813.2 1 561 131
          GenBank mRNA BC113955.1 - 1 560 131
          BC148217.1 - 1 556 131

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr698

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P312452 NM_146602.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P312452 BC113955.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P312452 NM_146602.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P312452 BC113955.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P114596 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P312452 NM_146602.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P312452 BC113955.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P312452 NM_146602.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P312452 BC113955.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P312452 NM_146602.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P312452 BC113955.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P312452 NM_146602.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P312452 BC113955.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396560 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396536 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P114596 - -
          Whole Mouse Genome G4122A A_51_P312452 NM_146602.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P312452 BC113955.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P312452 NM_146602.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P312452 BC113955.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P114596 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06219544 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06219547 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06219545 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06219541 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05962705 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06353964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06125633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396545 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05962721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396566 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06592717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05962703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396549 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566598 - -
          Affymetrix Mouse Gene 1.1 ST Array 10566597 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 530769 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 70452 - -
          Illumina mouse-6 v1.0 expression beadchip 70452 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000070452 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1213368 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1213368 - -
          mouse-6 v1.1 (Illumina) scl30861.2.98_141-S - -
          Illumina Sentrix mouse-6.v1 bead array 70452 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl30861.2.98_141-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 70452 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1213368 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1213368 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline