Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00837312_s1
          See other OLFR971 GE Assays ›
          Gene Symbol:
          Olfr971
          Gene Name:
          olfactory receptor 971
          Gene Aliases:
          MOR171-13
          Chromosome Location:
          Chr.9: 39839436 - 39840359 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00837312_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr971
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146614.1 NP_666825.1 1 49 119

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr971

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2057761 NM_146614.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156881 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2057761 NM_146614.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P472857 NM_146614.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440321 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440323 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440351 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440318 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440346 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440320 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156881 - -
          Whole Mouse Genome G4122A A_51_P472857 NM_146614.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106724 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156881 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07854876 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07854889 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08167616 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08167597 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07854868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440338 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08167616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440323 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440319 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440328 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08167610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440352 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08167605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08167597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07854871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440327 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10584513 - -
          Affymetrix Mouse Gene 1.1 ST Array 10584512 - -
          Affymetrix Mouse Clariom S Array PSR0900002912.mm.1 NM_146614.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002912.mm.1 NM_146614.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002912.mm.1 TC0900000439.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 585223 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 520685 - -
          Illumina mouse-6 v1.0 expression beadchip 520685 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000520685 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2729325 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2729325 - -
          mouse-6 v1.1 (Illumina) scl37075.1.12_21-S - -
          Illumina Sentrix mouse-6.v1 bead array 520685 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37075.1.12_21-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 520685 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2729325 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2729325 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline