Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00837332_s1
          See other OLFR292 GE Assays ›
          Gene Symbol:
          Olfr292
          Gene Name:
          olfactory receptor 292
          Gene Aliases:
          MOR220-2
          Chromosome Location:
          Chr.7: 86688341 - 86695384 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00837332_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr292
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146620.2 NP_666831.2 2 503 195

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr292

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P112532 NM_146620.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P112532 CB173961.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 855230 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 509643 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 20784 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 109918 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 177187 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171868 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126841 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P112532 NM_146620.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P112532 CB173961.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295482 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295465 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295475 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295460 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295479 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295463 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295477 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295468 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295480 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295472 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295474 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P126841 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112532 NM_146620.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112532 CB173961.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P126841 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06121149 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06121160 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06578093 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06578094 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06121148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295445 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06121156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06347007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295479 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295454 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295472 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06009832 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06063285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295452 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295480 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05899396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295469 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10554734 - -
          Affymetrix Mouse Gene 1.0 ST Array 10554735 - -
          Affymetrix Mouse Gene 1.1 ST Array 10554733 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 896044 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4150706 - -
          Illumina mouse-6 v1.0 expression beadchip 4150706 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004150706 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1231223 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1231223 - -
          mouse-6 v1.1 (Illumina) scl0258613.1_156-S - -
          Illumina Sentrix mouse-6.v1 bead array 4150706 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258613.1_156-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4150706 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1231223 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1231223 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0