Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00837358_s1
          See other OLFR120 GE Assays ›
          Gene Symbol:
          Olfr120
          Gene Name:
          olfactory receptor 120
          Gene Aliases:
          MOR263-3
          Chromosome Location:
          Chr.17: 37725999 - 37726991 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00837358_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr120
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146631.1 NP_666842.1 1 50 161

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr120

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P140200 NM_146631.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P140200 NM_146631.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P360658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169716 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169710 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169730 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169714 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169713 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169725 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169729 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169711 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169708 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169715 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169718 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169731 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169696 - -
          Whole Mouse Genome G4122A A_51_P360658 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13116108 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13143084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169716 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169706 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169685 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13256950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169711 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169713 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169690 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169693 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13061728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13061729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169716 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13256961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13256970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169700 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169740 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169714 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169731 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169708 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13412946 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169696 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169718 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169691 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169707 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169730 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10445152 - -
          Affymetrix Mouse Gene 1.1 ST Array 10445151 - -
          Affymetrix Mouse Clariom S Array PSR1700007733.mm.1 NM_146631.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007733.mm.1 NM_146631.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007733.mm.1 TC1700000780.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104920161 - -
          Illumina mouse-6 v1.0 expression beadchip 1170044 - -
          Illumina mouse-6 v1.0 expression beadchip 104920161 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001170044 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1242335 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1242335 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2535171 - -
          mouse-6 v1.1 (Illumina) GI_33238907-S - -
          mouse-6 v1.1 (Illumina) scl0258624.1_197-S - -
          Illumina Sentrix mouse-6.v1 bead array 104920161 - -
          Illumina Sentrix mouse-6.v1 bead array 1170044 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258624.1_197-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104920161 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1170044 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1242335 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2535171 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1242335 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline