Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00837400_s1
          See other OLFR1487 GE Assays ›
          Gene Symbol:
          Olfr1487
          Gene Name:
          olfactory receptor 1487
          Gene Aliases:
          MOR202-18
          Chromosome Location:
          Chr.19: 13619164 - 13620111 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00837400_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1487
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146636.1 NP_666847.1 1 445 173

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1487

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P618947 NM_146636.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P618947 NM_146636.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P618947 NM_146636.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P618947 NM_146636.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P618947 NM_146636.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P618947 NM_146636.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965198 - -
          Whole Mouse Genome G4122A A_52_P618947 NM_146636.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P618947 NM_146636.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114421 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117674 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14216142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965247 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14256124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14187349 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965224 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965205 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10461819 - -
          Affymetrix Mouse Gene 1.1 ST Array 10461818 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 786796 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6980341 - -
          Illumina mouse-6 v1.0 expression beadchip 100770594 - -
          Illumina mouse-6 v1.0 expression beadchip 6980341 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006980341 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2717136 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2717136 - -
          mouse-6 v1.1 (Illumina) GI_33239127-S - -
          mouse-6 v1.1 (Illumina) scl0258629.1_152-S - -
          Illumina Sentrix mouse-6.v1 bead array 6980341 - -
          Illumina Sentrix mouse-6.v1 bead array 100770594 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258629.1_152-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6980341 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100770594 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2717136 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3004676 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2717136 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3004676 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0