Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00837475_s1
          See other OLFR444 GE Assays ›
          Gene Symbol:
          Olfr444
          Gene Name:
          olfactory receptor 444
          Gene Aliases:
          MOR261-2
          Chromosome Location:
          Chr.6: 42955500 - 42956432 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00837475_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr444
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146656.1 NP_666867.1 1 454 82
          GenBank mRNA BC145664.1 - 1 495 40130967 82

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr444

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P597545 BC145664.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P597545 NM_146656.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P597545 BC145664.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P597545 NM_146656.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 213949 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P175611 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P597545 BC145664.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P597545 NM_146656.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P597545 BC145664.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P597545 NM_146656.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P597545 BC145664.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P597545 NM_146656.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P597545 BC145664.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P597545 NM_146656.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P597545 BC145664.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P597545 NM_146656.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P597545 BC145664.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P597545 NM_146656.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101790 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05130040 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05271781 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05271779 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05130046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05824669 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10537810 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537818 - -
          Affymetrix Mouse Gene 1.1 ST Array 10537809 - -
          Affymetrix Mouse Gene 1.1 ST Array 10537817 - -
          Affymetrix Mouse Clariom S Array PSR0600003746.mm.1 TC0600000474.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600003745.mm.1 TC0600000474.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600003745.mm.1 NM_146656.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600003745.mm.1 NM_146656.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600003744.mm.1 TC0600000474.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 387037 - -
          ABI Mouse Genome Survey Microarray 673010 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4060504 - -
          Illumina mouse-6 v1.0 expression beadchip 4060504 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004060504 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2589875 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2589875 - -
          mouse-6 v1.1 (Illumina) scl0258650.1_51-S - -
          Illumina Sentrix mouse-6.v1 bead array 4060504 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258650.1_51-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4060504 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2589875 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2873422 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2589875 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2873422 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline