Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00837515_s1
          See other OLFR740 GE Assays ›
          Gene Symbol:
          Olfr740
          Gene Name:
          olfactory receptor 740
          Gene Aliases:
          MOR106-4
          Chromosome Location:
          Chr.14: 50453054 - 50453989 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00837515_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr740
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146667.2 NP_666878.2 1 641 80

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr740

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2046220 NM_146667.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P169944 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2046220 NM_146667.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P390844 NM_146667.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597508 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29597499 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P169944 - -
          Whole Mouse Genome G4122A A_52_P390844 NM_146667.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125548 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P169944 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11583821 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11583808 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11583810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29597479 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11527049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597494 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11417336 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597496 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597528 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597545 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11417331 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597502 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597491 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11728126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597508 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597485 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597503 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597526 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597493 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597515 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11523648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597535 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29597548 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10414480 - -
          Affymetrix Mouse Gene 1.1 ST Array 10414479 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 899323 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 580278 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000580278 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2745719 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2745719 - -
          mouse-6 v1.1 (Illumina) scl46374.1.170_49-S - -
          Illumina Sentrix mouse-6.v1 bead array 580278 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl46374.1.170_49-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 580278 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2745719 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2745719 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0