Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00837599_s1
          See other OLFR1466 GE Assays ›
          Gene Symbol:
          Olfr1466
          Gene Name:
          olfactory receptor 1466
          Gene Aliases:
          MOR202-12
          Chromosome Location:
          Chr.19: 13338901 - 13342785 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00837599_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1466
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146694.1 NP_666905.1 1 449 77

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1466

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2175579 NM_146694.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118182 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2175579 NM_146694.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P215125 NM_146694.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31964181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31964159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31964185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31964146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31964179 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31964166 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31964164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31964163 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31964180 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31964152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31964157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31964172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31964183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31964186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31964149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31964145 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31964150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31964148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31964156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31964151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31964177 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31964154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31964162 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P118182 - -
          Whole Mouse Genome G4122A A_51_P215125 NM_146694.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116996 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P118182 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14251461 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14251460 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14251457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31964156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31964179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31964205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31964145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31964157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31964151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31964177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31964193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31964159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31964191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31964166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31964148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31964185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31964171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31964187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31964206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31964209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31964146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31964150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31964201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31964195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31964197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31964180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31964162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31964210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31964152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31964172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31964154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31964163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31964183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964156 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14503162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14221621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14474585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964197 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964151 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964187 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964172 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964164 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964145 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14503166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964204 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31964179 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14474580 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10461801 - -
          Affymetrix Mouse Gene 1.1 ST Array 10461800 - -
          Affymetrix Mouse Clariom S Array PSR1900003033.mm.1 NM_146694.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003033.mm.1 NM_146694.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003033.mm.1 TC1900000269.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 357952 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1940082 - -
          Illumina mouse-6 v1.0 expression beadchip 1940082 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001940082 - -
          Illumina Sentrix mouse-6.v1 bead array 1940082 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl6287.1.1_21-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1940082 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2920992 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2920992 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline