Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00837649_s1
          See other OLFR410 GE Assays ›
          Gene Symbol:
          Olfr410
          Gene Name:
          olfactory receptor 410
          Gene Aliases:
          MOR255-5
          Chromosome Location:
          Chr.11: 74334282 - 74335229 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00837649_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr410
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146707.1 NP_666918.1 1 755 159

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr410

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P206461 NM_146707.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919933 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919934 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919959 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919950 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919916 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919957 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27919931 - -
          Whole Mouse Genome G4122A A_51_P206461 NM_146707.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09709159 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09182888 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09709141 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09709160 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09269894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27919943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09709154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919914 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919908 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09625824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919916 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09340665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09340670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09340675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27919970 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10388307 - -
          Affymetrix Mouse Gene 1.1 ST Array 10388306 - -
          Affymetrix Mouse Clariom S Array PSR1100029401.mm.1 NM_146707.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029401.mm.1 NM_146707.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029401.mm.1 TC1100003191.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 874433 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6290142 - -
          Illumina mouse-6 v1.0 expression beadchip 6290142 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006290142 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1225411 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1225411 - -
          mouse-6 v1.1 (Illumina) scl00258702.1_217-S - -
          Illumina Sentrix mouse-6.v1 bead array 6290142 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00258702.1_217-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6290142 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1225411 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1225411 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0