Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00837697_sH
          See other OLFR784 GE Assays ›
          Gene Symbol:
          Olfr784
          Gene Name:
          olfactory receptor 784
          Gene Aliases:
          MOR110-3
          Chromosome Location:
          Chr.10: 129387635 - 129388594 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00837697_sH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr784
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146729.1 NP_666940.1 1 851 173

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000001640.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr784

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P147802 NM_146729.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P147802 NM_146729.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P147802 NM_146729.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P147802 NM_146729.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P147802 NM_146729.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P147802 NM_146729.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545690 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545649 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27545653 - -
          Whole Mouse Genome G4122A A_51_P147802 NM_146729.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P147802 NM_146729.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107804 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104417 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08483707 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08911875 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08483708 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08483705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27545657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08794128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08483708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545679 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08794126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08483705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27545674 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10367492 - -
          Affymetrix Mouse Gene 1.1 ST Array 10367491 - -
          Affymetrix Mouse Clariom S Array PSR1000012033.mm.1 NM_146729.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000012033.mm.1 NM_146729.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000012033.mm.1 TC1000001640.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 503624 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6860136 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006860136 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2623022 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2623022 - -
          mouse-6 v1.1 (Illumina) scl0258724.1_28-S - -
          Illumina Sentrix mouse-6.v1 bead array 6860136 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258724.1_28-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6860136 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2623022 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2623022 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline