Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00837698_s1
          See other OLFR483 GE Assays ›
          Gene Symbol:
          Olfr483
          Gene Name:
          olfactory receptor 483
          Gene Aliases:
          MOR204-12
          Chromosome Location:
          Chr.7: 108103311 - 108104258 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00837698_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr483
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146735.1 NP_666946.1 1 890 196
          GenBank mRNA BC116910.1 - 1 922 40126229 196
          BC116912.1 - 1 922 40126231 196

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001592.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr483

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of smell
          response to stimulus
          detection of chemical stimulus involved in sensory perception of smell
          signal transducer activity
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2082499 NM_146735.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2082499 BC116910.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2082499 BC116912.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2082499 NM_146735.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2082499 BC116910.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2082499 BC116912.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P382608 NM_146735.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P382608 BC116910.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P382608 BC116912.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25402341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25402350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25402344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25402348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25402347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25402345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25402349 - -
          Whole Mouse Genome G4122A A_51_P382608 NM_146735.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P382608 BC116910.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P382608 BC116912.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25402296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06441996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402314 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06209181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06481646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06178023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402323 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06666820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06625592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402373 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06178021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06666821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402352 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402319 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06666818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25402350 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10556132 - -
          Affymetrix Mouse Gene 1.1 ST Array 10556131 - -
          Affymetrix Mouse Clariom S Array PSR0700013273.mm.1 TC0700001592.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700013271.mm.1 TC0700001592.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700013272.mm.1 TC0700001592.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700013272.mm.1 NM_146735.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700013272.mm.1 NM_146735.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 791230 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4010537 - -
          Illumina mouse-6 v1.0 expression beadchip 4010537 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004010537 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2590828 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2590828 - -
          mouse-6 v1.1 (Illumina) scl0258730.1_57-S - -
          Illumina Sentrix mouse-6.v1 bead array 4010537 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258730.1_57-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4010537 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2590828 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2590828 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline