Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00837710_s1
          See other OLFR494 GE Assays ›
          Gene Symbol:
          Olfr494
          Gene Name:
          olfactory receptor 494
          Gene Aliases:
          MOR204-10
          Chromosome Location:
          Chr.7: 108367492 - 108368436 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00837710_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr494
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146737.1 NP_666948.1 1 355 182

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr494

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of smell
          response to stimulus
          detection of chemical stimulus involved in sensory perception of smell
          signal transducer activity
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P253426 NM_146737.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P253426 NM_146737.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P253426 NM_146737.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P253426 NM_146737.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P253426 NM_146737.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P253426 NM_146737.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403180 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403162 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403179 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25403177 - -
          Whole Mouse Genome G4122A A_51_P253426 NM_146737.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P253426 NM_146737.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122912 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06037637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06547513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06547514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25403180 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10556140 - -
          Affymetrix Mouse Gene 1.1 ST Array 10556139 - -
          Affymetrix Mouse Clariom S Array PSR0700013276.mm.1 TC0700001594.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700013276.mm.1 NM_146737.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700013276.mm.1 NM_146737.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 846473 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102900433 - -
          Illumina mouse-6 v1.0 expression beadchip 102900433 - -
          Illumina mouse-6 v1.0 expression beadchip 6550500 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006550500 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2706613 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2706613 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2535125 - -
          mouse-6 v1.1 (Illumina) GI_22129366-S - -
          mouse-6 v1.1 (Illumina) scl32218.1.1_74-S - -
          Illumina Sentrix mouse-6.v1 bead array 102900433 - -
          Illumina Sentrix mouse-6.v1 bead array 6550500 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32218.1.1_74-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102900433 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6550500 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2706613 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2782683 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2535125 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2706613 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2782683 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0