Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00837776_s1
          See other OLFR1248 GE Assays ›
          Gene Symbol:
          Olfr1248
          Gene Name:
          olfactory receptor 1248
          Gene Aliases:
          MOR23110
          Chromosome Location:
          Chr.2: 89617168 - 89618245 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00837776_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1248
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146791.2 NP_667002.2 1 606 87
          GenBank mRNA BC150718.1 - 1 608 9087629 87

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1248

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P504735 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P504735 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P504735 - -
          Whole Mouse Genome G4122A A_51_P504735 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123415 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112005 NM_207568.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112005 BC125497.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112005 BC125499.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414290 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02033188 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01691472 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414365 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414363 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414370 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02064567 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414338 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414334 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484842 - -
          Affymetrix Mouse Gene 1.0 ST Array 10484838 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484841 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484837 - -
          Affymetrix Mouse Clariom S Array PSR0200034087.mm.1 NM_146791.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034087.mm.1 NM_146791.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034087.mm.1 TC0200004066.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200034088.mm.1 NM_146791.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034088.mm.1 NM_146791.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034088.mm.1 TC0200004066.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200034086.mm.1 NM_146791.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034086.mm.1 NM_146791.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034086.mm.1 TC0200004066.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106350070 - -
          MouseWG-6 v2 Expression BeadChip 510100 - -
          Illumina mouse-6 v1.0 expression beadchip 510100 - -
          Illumina mouse-6 v1.0 expression beadchip 106350070 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000510100 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2748733 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2748733 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1242432 - -
          mouse-6 v1.1 (Illumina) scl0258787.1_44-S - -
          mouse-6 v1.1 (Illumina) GI_22129266-S - -
          Illumina Sentrix mouse-6.v1 bead array 510100 - -
          Illumina Sentrix mouse-6.v1 bead array 106350070 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258787.1_44-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106350070 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 510100 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2748733 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1242432 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2748733 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0258787.1_44-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline