Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00838007_s1
          See other OLFR1280 GE Assays ›
          Gene Symbol:
          Olfr1280
          Gene Name:
          olfactory receptor 1280
          Gene Aliases:
          MOR248-1
          Chromosome Location:
          Chr.2: 111308466 - 111374131 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00838007_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1280
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146908.1 NP_667119.1 1 87 76
          GenBank mRNA BC119051.1 - 1 130 8733800 76
          BC119053.1 - 1 130 8733802 76

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1280

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1968095 BC119053.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1968095 BC119051.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1968095 NM_146908.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1968095 BC119053.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1968095 BC119051.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1968095 NM_146908.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P263983 BC119053.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P263983 BC119051.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P263983 NM_146908.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524835 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524843 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524832 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524812 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524841 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524833 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524845 - -
          Whole Mouse Genome G4122A A_51_P263983 BC119053.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P263983 BC119051.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P263983 NM_146908.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133215 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111204 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01336179 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01336183 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02278939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524825 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524866 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524836 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524816 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01336191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524827 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524846 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524864 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524834 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524835 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524832 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524841 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524848 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524840 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524865 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524833 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524823 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02278939 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10474498 - -
          Affymetrix Mouse Gene 1.0 ST Array 10474487 - -
          Affymetrix Mouse Gene 1.0 ST Array 10474486 - -
          Affymetrix Mouse Gene 1.1 ST Array 10474497 - -
          Affymetrix Mouse Gene 1.1 ST Array 10474485 - -
          Affymetrix Mouse Clariom S Array PSR0200012261.mm.1 NM_146908.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200012261.mm.1 NM_146908.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200012261.mm.1 TC0200001642.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200012260.mm.1 TC0200001642.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 589042 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1400332 - -
          Illumina mouse-6 v1.0 expression beadchip 1400332 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001400332 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1255579 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1255579 - -
          mouse-6 v1.1 (Illumina) scl0258910.1_105-S - -
          Illumina Sentrix mouse-6.v1 bead array 1400332 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258910.1_105-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1400332 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1255579 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1255579 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline