Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00838033_s1
          See other OLFR20 GE Assays ›
          Gene Symbol:
          Olfr20
          Gene Name:
          olfactory receptor 20
          Gene Aliases:
          MOR135-11, MTPCR06, MTPCR55, Olfr21
          Chromosome Location:
          Chr.11: 73350859 - 73354699 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00838033_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr20
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146923.2 NP_667134.2 2 968 135
          GenBank mRNA BC138172.1 - 1 871 8861191 135
          BC138173.1 - 1 871 8861192 135

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr20

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1984941 NM_146923.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1984941 BC138172.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1984941 BC138173.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141381 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1984941 NM_146923.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1984941 BC138172.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1984941 BC138173.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P211150 NM_146923.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P211150 BC138172.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P211150 BC138173.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P211150 X89679.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P211150 X89692.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916772 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916753 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916731 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916729 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916730 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916777 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916775 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27916728 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141381 - -
          Whole Mouse Genome G4122A A_52_P211150 NM_146923.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P211150 BC138172.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P211150 BC138173.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P211150 X89679.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P211150 X89692.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138089 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128672 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137051 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141381 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09148697 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09148696 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09148701 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09479901 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09148698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09422909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09476543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09291139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916735 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916744 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916722 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916728 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916772 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09004494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916733 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10378398 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378396 - -
          Affymetrix Mouse Gene 1.1 ST Array 10378397 - -
          Affymetrix Mouse Gene 1.1 ST Array 10378395 - -
          Affymetrix Mouse Clariom S Array PSR1100009318.mm.1 TC1100001021.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 720721 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3290411 - -
          Illumina mouse-6 v1.0 expression beadchip 4920021 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004920021 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1215789 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215789 - -
          mouse-6 v1.1 (Illumina) scl00258925.1_327-S - -
          Illumina Sentrix mouse-6.v1 bead array 4920021 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00258925.1_327-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4920021 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2947005 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2947005 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline