Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00838056_s1
          See other OLFR802 GE Assays ›
          Gene Symbol:
          Olfr802
          Gene Name:
          olfactory receptor 802
          Gene Aliases:
          MOR111-1
          Chromosome Location:
          Chr.10: 129681799 - 129686098 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00838056_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr802
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146932.1 NP_667143.1 1 245 87

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr802

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957807 NM_146932.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121571 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P167610 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957807 NM_146932.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P276873 NM_146932.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546818 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546810 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546786 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546807 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546823 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546806 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546777 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546812 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546843 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546820 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546814 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546819 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546779 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546802 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546789 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546800 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546839 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546801 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546841 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546794 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546821 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546817 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27546775 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121571 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P167610 - -
          Whole Mouse Genome G4122A A_51_P276873 NM_146932.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108323 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121571 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P167610 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08477534 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08304650 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08304661 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08304653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27546820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08304661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08939122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08861979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08946667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546802 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546832 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08272459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08483274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27546781 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10373633 - -
          Affymetrix Mouse Gene 1.1 ST Array 10373632 - -
          Affymetrix Mouse Clariom S Array PSR1000023956.mm.1 NM_146932.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023956.mm.1 NM_146932.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023956.mm.1 TC1000003186.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 553001 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5550021 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005550021 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1218390 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1218390 - -
          mouse-6 v1.1 (Illumina) scl5160.1.1_61-S - -
          Illumina Sentrix mouse-6.v1 bead array 5550021 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl5160.1.1_61-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5550021 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1218390 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1218390 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline