Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00838177_s1
          See other OLFR176 GE Assays ›
          Gene Symbol:
          Olfr176
          Gene Name:
          olfactory receptor 176
          Gene Aliases:
          GA_x54KRFPKG5P-55043245-55042289, MOR184-8
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00838177_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr176
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146993.1 NP_667204.1 1 442 111

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr176

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2005595 NM_146993.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P74344 NM_146993.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P74344 NM_146993.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2005595 NM_146993.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P74344 NM_146993.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P74344 NM_146993.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P74344 NM_146993.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P74344 NM_146993.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30793591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30793588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30793589 - -
          Whole Mouse Genome G4122A A_52_P74344 NM_146993.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P74344 NM_146993.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12782935 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12983781 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12505893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793614 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12782953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793619 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793583 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10440140 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440142 - -
          Affymetrix Mouse Gene 1.1 ST Array 10440141 - -
          Affymetrix Mouse Gene 1.1 ST Array 10440138 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 902454 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3830301 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003830301 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2646904 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2646904 - -
          mouse-6 v1.1 (Illumina) scl00258995.1_310-S - -
          Illumina Sentrix mouse-6.v1 bead array 3830301 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00258995.1_310-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3830301 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2646904 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2646904 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline