Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00838194_s1
          See other OLFR178 GE Assays ›
          Gene Symbol:
          Olfr178
          Gene Name:
          olfactory receptor 178
          Gene Aliases:
          MOR184-6
          Chromosome Location:
          Chr.16: 58889259 - 58890218 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00838194_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr178
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146997.2 NP_667208.2 1 460 77
          GenBank mRNA BC111099.1 - 1 459 77
          BC127221.1 - 1 458 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600001783.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr178

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P399974 BC111099.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P399974 BC127221.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P399974 NM_146997.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P399974 BC111099.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P399974 BC127221.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P399974 NM_146997.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P399974 BC111099.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P399974 BC127221.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P399974 NM_146997.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P399974 BC111099.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P399974 BC127221.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P399974 NM_146997.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P399974 BC111099.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P399974 BC127221.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P399974 NM_146997.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P399974 BC111099.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P399974 BC127221.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P399974 NM_146997.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30793651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30793636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30793634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30793667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30793653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30793664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30793668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30793671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30793674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30793639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30793640 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30793660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30793657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30793658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30793642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30793662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30793637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30793638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30793672 - -
          Whole Mouse Genome G4122A A_51_P399974 BC111099.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P399974 BC127221.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P399974 NM_146997.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P399974 BC111099.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P399974 BC127221.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P399974 NM_146997.2 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12752387 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13024411 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13024400 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12752384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30793660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793629 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793671 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12752393 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793642 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793653 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12477680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793654 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793662 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30793674 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10440144 - -
          Affymetrix Mouse Gene 1.1 ST Array 10440143 - -
          Affymetrix Mouse Clariom S Array PSR1600014444.mm.1 NM_146997.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600014444.mm.1 NM_146997.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600014444.mm.1 TC1600001783.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 562616 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 7100332 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007100332 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1255012 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1255012 - -
          mouse-6 v1.1 (Illumina) scl480.1.1_326-S - -
          Illumina Sentrix mouse-6.v1 bead array 7100332 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl480.1.1_326-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7100332 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1255012 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1255012 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline