Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00838285_s1
          See other OLFR585 GE Assays ›
          Gene Symbol:
          Olfr585
          Gene Name:
          olfactory receptor 585
          Gene Aliases:
          MOR14-4
          Chromosome Location:
          Chr.7: 103097743 - 103098699 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00838285_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr585
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147087.2 NP_667298.2 1 648 94
          GenBank mRNA BC145897.1 - 1 40131200 94

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr585

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P375331 NM_147087.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P375331 BC145897.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P375331 NM_147087.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P375331 BC145897.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P375331 NM_147087.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P375331 BC145897.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P375331 NM_147087.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P375331 BC145897.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P375331 NM_147087.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P375331 BC145897.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P375331 NM_147087.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P375331 BC145897.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P375331 NM_147087.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P375331 BC145897.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P375331 NM_147087.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P375331 BC145897.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06591784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06085000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06575247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381983 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555761 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555765 - -
          Affymetrix Mouse Gene 1.1 ST Array 10555760 - -
          Affymetrix Mouse Gene 1.1 ST Array 10555764 - -
          Affymetrix Mouse Clariom S Array PSR0700012718.mm.1 NM_147087.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012718.mm.1 NM_147087.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012718.mm.1 TC0700001477.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012719.mm.1 TC0700001477.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2946430 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2946430 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline