Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00838306_s1
          See other OLFR980 GE Assays ›
          Gene Symbol:
          Olfr980
          Gene Name:
          olfactory receptor 980
          Gene Aliases:
          MOR223-2
          Chromosome Location:
          Chr.9: 40005994 - 40007029 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00838306_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr980
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147106.2 NP_667317.1 1 629 125
          GenBank mRNA BC116814.1 - 1 629 40126133 125
          BC119187.1 - 1 629 8733936 125

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr980

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1962541 BC119187.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1962541 NM_147106.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1962541 BC116814.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P115699 BC119187.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P115699 NM_147106.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P115699 BC116814.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147231 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P115699 BC119187.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P115699 NM_147106.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P115699 BC116814.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1962541 BC119187.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1962541 NM_147106.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1962541 BC116814.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P594854 BC119187.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P594854 NM_147106.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P594854 BC116814.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440958 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440972 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440934 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440947 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440976 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440967 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440950 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440977 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P147231 - -
          Whole Mouse Genome G4122A A_52_P594854 BC119187.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P594854 NM_147106.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P594854 BC116814.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115699 BC119187.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115699 NM_147106.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115699 BC116814.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P147231 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07925693 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07925705 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07925688 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07925712 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07925699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07615177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440986 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10592464 - -
          Affymetrix Mouse Gene 1.1 ST Array 10592463 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 899902 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4850075 - -
          Illumina mouse-6 v1.0 expression beadchip 4850075 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004850075 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1219492 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1219492 - -
          mouse-6 v1.1 (Illumina) scl0259110.1_3-S - -
          Illumina Sentrix mouse-6.v1 bead array 4850075 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0259110.1_3-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4850075 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1219492 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1219492 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0