Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00838384_g1
          See other EVA1B GE Assays ›
          Gene Symbol:
          Eva1b
          Gene Name:
          eva-1 homolog B (C. elegans)
          Gene Aliases:
          2610027C15Rik, Fam176b
          Chromosome Location:
          Chr.4: 126145704 - 126149879 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00838384_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eva1b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172145.3 NP_742157.1 2 - 3 725 249
          GenBank mRNA AK076006.1 - 2 - 3 727 249
          AK077984.1 - 2 - 3 1053 251
          BC029204.1 - 1 - 2 358 3964022 249

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001455.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Eva1b

          Gene Ontology Categories:

          Function(s) Process(es)

          mitophagy in response to mitochondrial depolarization
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23278480 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23278479 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23278481 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23278482 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23278470 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23278476 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23278473 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23278478 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23278471 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23278477 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23278475 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2087796 BC029204.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2087796 AK076006.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2087796 NM_172145.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P678056 AK076006.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P678056 NM_172145.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P678056 AK076006.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P678056 NM_172145.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 181716 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153933 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P678056 AK076006.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P678056 NM_172145.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P678056 AK076006.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P678056 NM_172145.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2087796 BC029204.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2087796 AK076006.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2087796 NM_172145.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23278483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23278470 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23278464 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23278484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23278448 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23278452 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23278441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23278473 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23278446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23278478 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23278474 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23278450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23278465 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23278443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23278456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23278467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23278451 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23278438 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23278485 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23278468 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23278482 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23278476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23278459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23278460 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23278463 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153933 - -
          Whole Mouse Genome G4122A A_52_P678056 AK076006.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P678056 NM_172145.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P678056 AK076006.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P678056 NM_172145.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P381506 AK077984.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P381506 AK076006.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P381506 NM_172145.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130983 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153933 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03556471 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03935172 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04025730 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04025731 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04025728 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23278470 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23278471 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23278480 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23278479 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23278475 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23278476 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23278481 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23278473 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23278482 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23278477 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23278478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23278436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23278476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23278444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23278484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23278482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23278441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23278435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23278433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23278473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23278456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23278483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23278485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23278526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23278431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23278470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23278522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23278520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23278474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23278463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23278452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23278478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23278467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23278438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23278465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23278450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23278519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23278439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23278459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23278448 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23278461 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03935202 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23278476 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23278444 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23278483 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23278452 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03512322 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23278431 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03512317 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23278463 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23278456 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23278433 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23278448 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23278473 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23278450 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23278469 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23278435 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23278447 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03688813 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03862925 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23278468 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03306322 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23278436 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23278460 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03935175 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03935160 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03890296 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03232838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23278459 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03935181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23278434 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04025730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23278443 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23278465 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10508130 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508131 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508132 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508133 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508134 - -
          Affymetrix HT MG-430 PM Array Plate 1454933_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10508128 - -
          Mouse Genome 430 2.0 Array 1454933_at - -
          Mouse Genome 430 2.0 Array 1454933_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 230752_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 230752_at - -
          Affymetrix Mouse Clariom S Array PSR0400011097.mm.1 TC0400001455.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011098.mm.1 TC0400001455.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011100.mm.1 NM_172145.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011100.mm.1 NM_172145.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011100.mm.1 TC0400001455.mm.1 MTA TC
          Mouse Expression Set 430 1454933_at - -
          Mouse Expression Set 430 1454933_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1454933_at - -
          Affymetrix HT Mouse Genome 430B Array 1454933_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 711411 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6220035 - -
          Illumina mouse-6 v1.0 expression beadchip 6220035 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006220035 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2714073 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2714073 - -
          mouse-6 v1.1 (Illumina) scl24968.2_41-S - -
          Illumina Sentrix mouse-6.v1 bead array 6220035 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl24968.2_41-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6220035 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2714073 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2890357 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2890357 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2714073 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5rdcx:80/100.66.79.29:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline