Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00838638_s1
          See other HIST1H3C GE Assays ›
          Gene Symbol:
          Hist1h3c
          Gene Name:
          histone cluster 1, H3c
          Gene Aliases:
          H3-143
          Chromosome Location:
          Chr.13: 23745042 - 23745521 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00838638_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hist1h3c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175653.2 NP_783584.1 1 355 132
          GenBank mRNA BC120800.1 - 1 436 40129723 132
          BC120802.1 - 1 436 40129725 132

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300001642.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hist1h3c

          Panther Classification:

          Molecular Function -

          histone

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleosome assembly
          histone binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28883243 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28883256 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28883258 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2145401 BC120800.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2145401 BC120802.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2145401 NM_175653.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2145401 BC120800.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2145401 BC120802.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2145401 NM_175653.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883253 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883250 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883251 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883248 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121737 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11047424 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11047420 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11047398 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11047413 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11047419 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11047422 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28883243 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28883256 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28883258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883323 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883241 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883236 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10511374 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883248 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883228 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883251 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28883225 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10404066 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408240 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408203 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408248 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404050 - -
          Affymetrix Mouse Gene 1.1 ST Array 10404065 - -
          Affymetrix Mouse Gene 1.1 ST Array 10408239 - -
          Affymetrix Mouse Clariom S Array PSR1300010186.mm.1 TC1300001642.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300010183.mm.1 TC1300001642.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300010184.mm.1 NM_175653.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010184.mm.1 TC1300001642.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300010182.mm.1 TC1300001642.mm.1 MTA TC
          Mouse Expression Set 430 1460314_s_at - -
          Mouse Expression Set 430 1418366_PM_at - -
          Mouse Expression Set 430 1418367_x_at - -
          Mouse Expression Set 430 1427864_PM_at - -
          Mouse Expression Set 430 1418367_PM_x_at - -
          Mouse Expression Set 430 1453573_PM_at - -
          Mouse Expression Set 430 1418366_at - -
          Mouse Expression Set 430 1427864_at - -
          Mouse Expression Set 430 1460314_PM_s_at - -
          Mouse Expression Set 430 1453573_at - -
          Mouse Expression Set 430 1442051_PM_at - -
          Mouse Expression Set 430 1442051_at - -
          Murine Genome U74v2 Set 93022_i_at - -
          Murine Genome U74v2 Set 94756_PM_at - -
          Murine Genome U74v2 Set 96416_f_at - -
          Murine Genome U74v2 Set 93023_PM_f_at - -
          Murine Genome U74v2 Set 93022_PM_i_at - -
          Murine Genome U74v2 Set 94756_at - -
          Murine Genome U74v2 Set 96416_PM_f_at - -
          Murine Genome U74v2 Set 93068_r_at - -
          Murine Genome U74v2 Set 93067_f_at - -
          Murine Genome U74v2 Set 93067_PM_f_at - -
          Murine Genome U74v2 Set 93068_PM_r_at - -
          Murine Genome U74v2 Set 93023_f_at - -
          Mouse Genome 430A 2.0 Array 1427864_PM_at - -
          Mouse Genome 430A 2.0 Array 1427864_at - -
          Mouse Genome 430A 2.0 Array 1453573_at - -
          Mouse Genome 430A 2.0 Array 1418367_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1418366_at - -
          Mouse Genome 430A 2.0 Array 1418366_PM_at - -
          Mouse Genome 430A 2.0 Array 1418367_x_at - -
          Mouse Genome 430A 2.0 Array 1453573_PM_at - -
          Mouse Genome 430A 2.0 Array 1460314_s_at - -
          Mouse Genome 430A 2.0 Array 1460314_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 730204 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000730204 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1229755 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1229755 - -
          mouse-6 v1.1 (Illumina) scl11507.1.1_324-S - -
          Illumina Sentrix mouse-6.v1 bead array 730204 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl11507.1.1_324-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 730204 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1229755 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1229755 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline